Product Image
Invitrogen™

pcDNA™3.1 Directional TOPO™ Expression Kit

The pcDNA™3.1 Directional TOPO™ Expression Kit uses linearized, topoisomerase I-activated pcDNA3.1D/V5-His-TOPO™ for five-minute directional cloning and subsequent high-level expression.Directional CloningRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
K49004040 Reactions
K49000120 Reactions
Catalog number K490040
Price (USD)
2,380.00
Each
Quantity:
40 Reactions
Price (USD)
2,380.00
Each
The pcDNA™3.1 Directional TOPO™ Expression Kit uses linearized, topoisomerase I-activated pcDNA3.1D/V5-His-TOPO™ for five-minute directional cloning and subsequent high-level expression.

Directional Cloning technology facilitates expression experiments because:

• A proofreading enzyme is used for fewer errors in cloned genes
• Greater than 90% of the clones are in the correct orientation for expression, reducing time spent colony screening for clone orientation

In addition, pcDNA3.1D/V5-His-TOPO™ provides strong expression levels from the CMV promoter and the option of a C-terminal V5-His fusion tag for easy detection of recombinant protein with an Anti-V5 Antibody and rapid purification on nickel-chelating resin.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
For Use With (Application)Constitutive Expression
Product TypeTOPO Expression Kit
Quantity40 Reactions
Selection Agent (Eukaryotic)Geneticin™ (G-418)
VectorDirectional TOPO Vectors, pcDNA
Cloning MethodDirectional TOPO
Product LineTOPO, pcDNA
PromoterCMV
Protein TagHis Tag (6x), V5 Epitope Tag
Unit SizeEach
Contents & Storage
2 × Box 1 (Store at -20°C):
• 20 μl pcDNA™3.1D/V5-His-TOPO™ (15–20 ng/μl)
• 10 μl dNTP Mix
• 50 μl Salt Solution
• 1 ml Sterile Water
• 20 μl T7 Sequencing Primer (0.1 μg/μl)
• 20 μl BGH Reverse Sequencing Primer (0.1 μg/μl)
• 10 μl Control PCR Template (0.1 μg/μl)
• 10 μl Control PCR Primers (0.1 μg/μl each)
• 10 μl Expression Plasmid (0.5 μg/μl)

2 × Box 2 (Store at -80°C):
• 6 ml S.O.C. Medium (may be stored at 4°C or room temperature)
• 21 × 50 μl TOP10 Cells
• 50 μl pUC19 Control DNA

Frequently asked questions (FAQs)

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

What is the difference between pcDNA3.1 vectors and the pcDNA3.3-TOPO vector?

pcDNA3.1 vectors contain the core CMV promoter that is truncated before the start of transcription, whereas the pcDNA 3.3-TOPO vector has the 672 bp native CMV promoter. This native CMV promoter allows high-level gene expression with two- to five-fold higher protein yields compared to other expression vectors. pcDNA3.1 vectors are available in restriction, TOPO, and Gateway cloning versions and as untagged and epitope-tagged versions, whereas the pcDNA3.3-TOPO vector is a TOPO TA-adapted, untagged vector that can be used to express native proteins without extraneous amino acids, and is hence ideal for antibody production and structural biology.

Citations & References (3)

Citations & References
Abstract
Functional analysis of the human papillomavirus type 16 E1=E4 protein provides a mechanism for in vivo and in vitro keratin filament reorganization.
Authors:Wang Q, Griffin H, Southern S, Jackson D, Martin A, McIntosh P, Davy C, Masterson PJ, Walker PA, Laskey P, Omary MB, Doorbar J,
Journal:J Virol
PubMed ID:14694114
'High-risk human papillomaviruses, such as human papillomavirus type 16 (HPV16), are the primary cause of cervical cancer. The HPV16 E1=E4 protein associates with keratin intermediate filaments and causes network collapse when expressed in epithelial cells in vitro. Here, we show that keratin association and network reorganization also occur in vivo ... More
Kaposi's sarcoma-associated herpesvirus K8 exon 3 contains three 5'-splice sites and harbors a K8.1 transcription start site.
Authors: Tang Shuang; Zheng Zhi-Ming;
Journal:J Biol Chem
PubMed ID:11832484
Kaposi's sarcoma-associated herpesvirus (KSHV) K8 and K8.1 open reading frames are juxtaposed and span from nucleotide (nt) 74850 to 76695 of the virus genome. A K8 pre-mRNA overlaps the entire K8.1 coding region, and alternative splicing of KSHV K8 and K8.1 pre-mRNAs each produces three isoforms (alpha, beta, and gamma) ... More
Blood group A glycosyltransferase occurring as alleles with high sequence difference is transiently induced during a Nippostrongylus brasiliensis parasite infection.
Authors: Olson Fredrik J; Johansson Malin E V; Klinga-Levan Karin; Bouhours Danièle; Enerbäck Lennart; Hansson Gunnar C; Karlsson Niclas G;
Journal:J Biol Chem
PubMed ID:11842091
Neutral mucin oligosaccharides from the small intestine of control rats and rats infected with the parasite Nippostrongylus brasiliensis were released and analyzed by gas chromatography-mass spectrometry. Infected animals expressed seven blood group A-like structures that were all absent in the control animals. The blood group A nature of these epitopes ... More