Product Image
Invitrogen™
Flp-In™ Core System
Our Flp-In™ Core Kit (part of the Flp-In™ Complete System) contains the necessary vectors that allow integration and expression ofRead more
Have Questions?
Catalog NumberQuantity
K601002
also known as K6010-02
1 kit
Catalog number K601002
also known as K6010-02
Price (USD)
1,894.00
Each
Quantity:
1 kit
Price (USD)
1,894.00
Each
Our Flp-In™ Core Kit (part of the Flp-In™ Complete System) contains the necessary vectors that allow integration and expression of your gene of interest in mammalian cells at a specific genomic location. The Flp-In System involves introduction of a Flp Recombination Target (FRT) site into the genome of the mammalian cell line of choice. An expression vector containing your gene of interest is then integrated into the genome via Flp recombinase-mediated DNA recombination at the FRT site.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
For Use With (Application)Targeted Integration
Product TypeFlp-In Core System
Quantity1 kit
Selection Agent (Eukaryotic)Zeocin™
VectorpFRT
Cloning MethodRestriction Enzyme/MCS
Product LineFlp-In, pcDNA
PromoterCMV
Protein TagUntagged
Unit SizeEach
Contents & Storage
The Flp-In™ Core Kit includes 20 μg each of pFRT⁄lacZeo, pOG44, pcDNA™5⁄FRT, and pcDNA™5⁄FRT⁄CAT (expression control), and 2 μg each of forward and reverse sequencing primers. Store at -20C.
Frequently asked questions (FAQs)
I performed the Flp-In reaction and obtained hygromycin-resistant clones which also turned out to be Zeocin antibiotic resistant and lacZ-positive. I am concerned that the Flp-In reaction has not worked. Can you explain what might be happening?
We have observed in-house that in cells where the FRT site has integrated into a very transcriptionally active locus in the host cell genome (seen more commonly in Flp-In CHO and Flp-In 293 cells but can also happen in Flp-In 3T3 cells and any other Flp-In host cell line), there is some "read-through" transcription and translation of the lacZ-Zeocin ORF subsequent to the Flp-In reaction, even though the lacZ-Zeocin ORF does not have a bonafide promoter and ATG. In such cases, the hygromycin-resistant clones would also be lacZ-positive and Zeocin antibiotic-resistant. To make sure that the integration is FRT site-specific and not random, we recommend doing a parallel control transfection with no pOG44 present. This should yield no surviving clones upon hygromycin selection, indicating that all the hygromycin-resistant clones obtained in the presence of pOG44 are indeed Flp recombinase-dependent and hence have the gene of interest integrated at the FRT site. Also, a Southern blot analysis of these clones will help verify that they do indeed have proper FRT integration of the gene of interest despite the expression of lacZ (although this is typically not necessary). After the Flp-In reaction, as long as you see hygromycin-resistant clones, we recommend that you select them and assay them for expression of your gene of interest.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
I used the pcDNA5/FRT vector in your Flp-In 3T3 cell line and was initially able to express my gene of interest, but after a month or so, lost expression. Why is this?
The Flp-In 3T3 cell line is derived from NIH3T3 cells, which are mouse fibroblast cells. The CMV promoter is known to get silenced over time in murine cell lines and hence we would recommend using a Flp-In expression vector with a non-CMV promoter in these cells, such as the pEF5/FRT/V5-D-TOPO vector or the pEF5/FRT/V5-DEST vector.
I used the pFRT/lacZeo vector to generate my own Flp-In host cell line and then made my Flp-In expression cell line. I am seeing very poor expression of my gene of interest. Is there anything else I can try to improve expression?
Before giving up, we would suggest that you try using the pFRT/lacZeo2 vector to generate your host cell line. This vector contains a truncated version of the SV40 promoter driving the lacZ-Zeocin fusion. Use of this vector facilitates the isolation of clones that have integrated the vector near enhancer elements in the genome, thus resulting in higher levels of expression of the gene of interest.
What is the difference between the Jump-In and Flp-In systems?
The Jump-In system is PhiC31-integrase mediated and is a stable, targeted, and irreversible mammalian expression system. It consists of the Jump-In Fast system that involves a single integration step and the Jump-InTI (targeted integration) system that needs two integration steps, both of which are targeted and irreversible. In contrast, the Flp-In system is a stable, targeted mammalian expression system that is reversible. The first integration is random (integration of pFRT/lacZeo), and the second integration (integration of the Flp-In expression vector) is targeted but reversible.
Is multiple integration of the Flp-In expression construct possible? How do you screen for multiple integrants, and how stable is the Flp-In expression cell line?
In theory, one can get multiple integrations of the Flp-In expression construct—an FRT-specific integration event and a random, second-site integration. However, random integration is a relatively uncommon event. Limiting the amount of DNA in the transfection will reduce the chance of second-site integration. We have transfected 293 cells (lacking the FRT site) with the pcDNA5/FRT vector and have identified one potential second-site integrant after screening over 200 clones. DNA integrations can be detected by Southern blot. A single integrant will display a single band; double: two; triple: three, etc. We have maintained a number of Flp-In expression cell lines for over four months and have not observed any loss of the Flp-In expression construct, whether hygromycin selection was maintained or not.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Citations & References (7)
Citations & References
Abstract
Genetic analysis of the herpes simplex virus type 1 UL20 protein domains involved in cytoplasmic virion envelopment and virus-induced cell fusion.
Authors:Melancon JM, Foster TP, Kousoulas KG,
Journal:J Virol
PubMed ID:15220406
'The herpes simplex virus type 1 UL20 protein (UL20p) is an important determinant for cytoplasmic virion morphogenesis and virus-induced cell fusion. To delineate the functional domains of the UL20 protein, we generated a panel of single and multiple (cluster) alanine substitutions as well as UL20p carboxyl-terminal truncations. The UL20 mutant ... More
Localization of the discontinuous immunodominant region recognized by human anti-thyroperoxidase autoantibodies in autoimmune thyroid diseases.
Authors:Bresson D, Cerutti M, Devauchelle G, Pugnière M, Roquet F, Bes C, Bossard C, Chardès T, Péraldi-Roux S,
Journal:J Biol Chem
PubMed ID:12501244
'The discontinuous immunodominant region (IDR) recognized by autoantibodies directed against the thyroperoxidase (TPO) molecule, a major autoantigen in autoimmune thyroid diseases, has not yet been completely localized. By using peptide phage-displayed technology, we identified three critical motifs, LXPEXD, QSYP, and EX(E/D)PPV, within selected mimotopes which interacted with the human recombinant ... More
GlcNAc 2-epimerase can serve a catabolic role in sialic acid metabolism.
Authors:Luchansky SJ, Yarema KJ, Takahashi S, Bertozzi CR,
Journal:J Biol Chem
PubMed ID:12499362
'Sialic acid is a major determinant of carbohydrate-receptor interactions in many systems pertinent to human health and disease. N-Acetylmannosamine (ManNAc) is the first committed intermediate in the sialic acid biosynthetic pathway; thus, the mechanisms that control intracellular ManNAc levels are important regulators of sialic acid production. UDP-GlcNAc 2-epimerase and GlcNAc ... More
Stable transmission of targeted gene modification using single-stranded oligonucleotides with flanking LNAs.
Authors:Andrieu-Soler C, Casas M, Faussat AM, Gandolphe C, Doat M, Tempé D, Giovannangeli C, Behar-Cohen F, Concordet JP,
Journal:Nucleic Acids Res
PubMed ID:16002788
Targeted mutagenesis directed by oligonucleotides (ONs) is a promising method for manipulating the genome in higher eukaryotes. In this study, we have compared gene editing by different ONs on two new target sequences, the eBFP and the rd1 mutant photoreceptor betaPDE cDNAs, which were integrated as single copy transgenes at ... More
Mevalonate kinase is a cytosolic enzyme in humans.
Authors:Hogenboom S, Tuyp JJ, Espeel M, Koster J, Wanders RJ, Waterham HR,
Journal:J Cell Sci
PubMed ID:14730012
In the past decade several reports have appeared which suggest that peroxisomes play a central role in isoprenoid/cholesterol biosynthesis. These suggestions were based primarily on the reported finding of several of the enzymes of the presqualene segment of the biosynthetic pathway in peroxisomes. More recently, however, conflicting results have been ... More