InsectSelect™ Glow Kit with Sf9 Cells - FAQs
InsectSelect™ Glow Kit with Sf9 Cells - FAQs
View additional product information for InsectSelect™ Glow Kit with Sf9 Cells - FAQs (K81001)
1 product FAQs found
Does the incubator need to be humidified when culturing insect cells?
Humidification is only necessary for monolayer cultures and suspension cultures with volumes less than 35 mL. With shaker or spinner cultures greater than 35 mL, humidification is not necessary.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.