FreeStyle™ MAX CHO Expression System - FAQs
FreeStyle™ MAX CHO Expression System - FAQs
View additional product information for FreeStyle™ MAX CHO Expression System - FAQs (K900020)
2 product FAQs found
I am interested in a mammalian expression system that will allow me to obtain very high yields of my protein. What kind of systems do you offer?
For stable high-yield expression, we offer the Freedom CHO-S and Freedom DG44 kits. For transient high-yield expression, we offer the ExpiCHO Expression System, Expi293 Expression System, FreeStyle 293 Expression System, FreeStyle Max 293 Expression System, and FreeStyle Max CHO Expression System. For high-yield expression of functional membrane proteins in Exp293F cells, we offer the Expi293 MembranePro Expression System (Cat. Nos. A25869, A25870) that combines the scalability and ease of use of Expi293 and the technology of MembranePro to allow an increase of more than 20-fold in membrane protein yield compared to the standard, adherent culture MembranePro Functional Protein Expression System. Please see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/ProteinExpressionAnalysis/pdfs/PG1391-PJ6257-CO28704-Expi293Adaption-MembraneProTech_AppNoteGlobal-FHR.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What vector do you recommend using with FreeSytle MAX CHO Expression System (Cat. No. K900020)?
We recommend using the pcDNA 3.4 TOPO TA Cloning Kit (Cat. No. A14697).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.