APO BrdU (TUNEL) Kit
Invitrogen™

APO BrdU (TUNEL) Kit

The ApoTarget™ APO-BRDU Kit is a two-color staining method for labeling DNA breaks and total cellular DNA to detect apoptoticRead more
Have Questions?
Catalog NumberQuantity
KHO100160 tests
Catalog number KHO1001
Price (USD)
924.00
Each
Quantity:
60 tests
Price (USD)
924.00
Each
The ApoTarget™ APO-BRDU Kit is a two-color staining method for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry.

View a selection guide for all apoptosis assays for flow cytometry.
Specifications
ColorGreen, Red
Excitation/EmissionFITC: 490/525
PI: 535/617
Flow Cytometer Laser Lines488
For Use With (Application)Flow Cytometry
For Use With (Equipment)Flow Cytometer
Label TypeClassic Dyes
Label or DyeFITC (Fluorescein), PI (Propidium Iodide)
No. of Reactions60
Product LineApoTarget
Product TypeApo-BRDU Tunel Kit
Quantity60 tests
Detection MethodFluorescence
FormatTube
Unit SizeEach
Contents & Storage
Contains pre-coated plate, standard, detector antibody, diluents, wash buffer, chromogen, stop solution and complete protocol

Citations & References (8)

Citations & References
Abstract
DNA damage induced by metronidazole in Giardia duodenalis triggers a DNA homologous recombination response.
Authors:Ordoñez-Quiroz A, Ortega-Pierres MG, Bazán-Tejeda ML, Bermúdez-Cruz RM
Journal:Exp Parasitol
PubMed ID:30237050
'The mechanisms underlying metronidazole (MTZ) resistance in Giardia duodenalis have been associated with decreased activity of the enzymes implicated in its activation including nitroductase-1, thioredoxin reductase and pyruvate-ferredoxin oxidoreductase (PFOR). MTZ activation generates radicals that can form adducts with proteins such as thioredoxin reductase and a- and -ß giardins as ... More
The effect of the melatonin on cryopreserved mouse testicular cells.
Authors:Saki G, Mirhoseini M, Hemadi M, Khodadadi A, Beygom Talebpour Amiri F
Journal:Int J Reprod Biomed
PubMed ID:27141545
'After improvements in various cancer treatments, life expectancy has been raised, but success in treatment causes loss of fertility in many of the survived young men. Cryopreservation of immature testicular tissues or cells introduced as the only way to preserve fertility. However, freezing has some harmful effects. Melatonin, a pineal ... More
Inhibition of MAPKinase pathway sensitizes thyroid cancer cells to ABT-737 induced apoptosis.
Authors:Gunda V, Sarosiek KA, Brauner E, Kim YS, Amin S, Zhou Z, Letai A, Parangi S
Journal:Cancer Lett
PubMed ID:28259821
'Bcl2 family proteins play an important role in the resistance of thyroid cancer cells to apoptosis induced by chemotherapeutic drugs and targeted therapies. BH3-profiling of seven fresh primary papillary thyroid cancer (PTC) tumors showed dependence for survival on Bcl-xL (2/7), Bcl2 (2/7), and Mcl-1 (2/7), while the majority of thyroid ... More
Disruption of galectin-3 and galectin-3 binding protein (G3BP) interaction by dietary pectic polysaccharides (DPP) - Arrest of metastasis, inhibition of proliferation and induction of apoptosis.
Authors:Natarajamurthy SH, Sistla S, Dharmesh SM
Journal:Int J Biol Macromol
PubMed ID:31356934
'Galectin-3 and galectin-3 binding proteins (G3BP) are implicated as key players in metastasis. In the current study, we evaluated the effect of pectic polysaccharides on galectin-3 and G3BP mediated metastasis in vitro (cells) and in vivo (tissues). In vitro study (double immunostaining) confirms the presence of galectin-3 on the cell ... More
Characterization of the cell-free DNA released by cultured cancer cells.
Authors:Bronkhorst AJ, Wentzel JF, Aucamp J, van Dyk E, du Plessis L, Pretorius PJ
Journal:Biochim Biophys Acta
PubMed ID:26529550
'The most prominent factor that delays the translation of cell-free DNA (cfDNA) analyses to clinical practice is the lack of knowledge regarding its origin and composition. The elucidation of the former is complicated by the seemingly random fluctuation of quantitative and qualitative characteristics of cfDNA in the blood of healthy ... More