Human NGAL ELISA Kit - FAQs
Human NGAL ELISA Kit - FAQs
View additional product information for Human NGAL ELISA Kit - FAQs (KIT036)
5 product FAQs found
Can you provide the shelf life for NGAL Human ELISA Kit (Cat. No. KIT036)?
NGAL Human ELISA Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (between 2-8 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).