L-Glutamic acid dimethyl ester hydrochloride, 98+%
L-Glutamic acid dimethyl ester hydrochloride, 98+%
L-Glutamic acid dimethyl ester hydrochloride, 98+%
Thermo Scientific Chemicals

L-Glutamic acid dimethyl ester hydrochloride, 98+%

CAS: 23150-65-4 | C7H13ClNO4 | 210.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L03764.06
also known as L03764-06
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.60
Save 6.95
Each
Chemical Identifiers
CAS23150-65-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)≤2%
Optical Rotation+25° to +27° (c=5 in water)
L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

Solubility
Soluble in water, methanol (50 mg/ml).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture, heat.
RUO – Research Use Only

General References:

  1. Stone T. W.Cortical pyramidal tract interneurones and their sensitivity to L-glutamic acid. The Journal of Physiology. 1973, 233, (1), 211-225.
  2. James S. Nowick; Noel A. Powell; Tram M. Nguyen; Glenn Noronha. An improved method for the synthesis of enantiomerically pure amino acid ester isocyanates. J. Org. Chem. 1992, 57, (26), 7364-7366.