5-Aminouracil, 97%
5-Aminouracil, 97%
5-Aminouracil, 97%
Thermo Scientific Chemicals

5-Aminouracil, 97%

CAS: 932-52-5 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L04452.06
also known as L04452-06
Price (USD)
35.65
Online Exclusive
38.90
Save 3.25 (8%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
38.90
Save 3.25 (8%)
Each
Chemical Identifiers
CAS932-52-5
IUPAC Name5-amino-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H5N3O2
InChI KeyBISHACNKZIBDFM-UHFFFAOYSA-N
SMILESNC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to cream to yellow to orange to brown
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only