delta-Valerolactone, 98%, may contain polymer - FAQs
delta-Valerolactone, 98%, may contain polymer - FAQs
View additional product information for delta-Valerolactone, 98%, may contain polymer - FAQs (L04794.14, L04794.36, L04794.22)
1 product FAQs found
What should I do if delta-Valerolactone, 98% (Cat. No. L04794.14, L04794.22, L04794.36) arrives with polymerization?
The quickest and simplest way to remove the polymeric material in the product is by filtering it through a pad of Celite under nitrogen.