4-Fluoro-DL-phenylalanine, 98+%
4-Fluoro-DL-phenylalanine, 98+%
4-Fluoro-DL-phenylalanine, 98+%
Thermo Scientific Chemicals

4-Fluoro-DL-phenylalanine, 98+%

CAS: 51-65-0 | C9H10FNO2 | 183.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07585.03
also known as L07585-03
Price (USD)
43.65
Online Exclusive
48.30
Save 4.65 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.30
Save 4.65 (10%)
Each
Chemical Identifiers
CAS51-65-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (HPLC)≥97.5% (Non-U.S. Specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (Non-U.S. specification)
Proton NMRConform to structure (U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only