beta-D-Glucosamine pentaacetate, 96%
beta-D-Glucosamine pentaacetate, 96%
beta-D-Glucosamine pentaacetate, 96%
Thermo Scientific Chemicals

beta-D-Glucosamine pentaacetate, 96%

CAS: 7772-79-4 | C16H23NO10 | 389.357 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09020.06
also known as L09020-06
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS7772-79-4
IUPAC Name[3,4,6-tris(acetyloxy)-5-acetamidooxan-2-yl]methyl acetate
Molecular FormulaC16H23NO10
InChI KeyOVPIZHVSWNOZMN-UHFFFAOYNA-N
SMILESCC(=O)NC1C(OC(C)=O)OC(COC(C)=O)C(OC(C)=O)C1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥95.0%
Optical Rotation1.5 ± 0.2? (C=1 in chloroform)
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)181.0-192.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only