N-Boc-sarcosine, 99%
N-Boc-sarcosine, 99%
N-Boc-sarcosine, 99%
Thermo Scientific Chemicals

N-Boc-sarcosine, 99%

CAS: 13734-36-6 | C8H15NO4 | 189.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13482.06
also known as L13482-06
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.80
Save 7.15 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.80
Save 7.15 (15%)
Each
Chemical Identifiers
CAS13734-36-6
IUPAC Name2-{[(tert-butoxy)carbonyl](methyl)amino}acetate
Molecular FormulaC8H14NO4
InChI KeyYRXIMPFOTQVOHG-UHFFFAOYSA-M
SMILESCN(CC([O-])=O)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only