2'-Deoxycytidine hydrochloride, 99%
2'-Deoxycytidine hydrochloride, 99%
2'-Deoxycytidine hydrochloride, 99%
Thermo Scientific Chemicals

2'-Deoxycytidine hydrochloride, 99%

CAS: 3992-42-5 | C9H14ClN3O4 | 263.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L14153.MD
also known as L14153-MD
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.40
Save 3.75 (13%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
24.65
Special offer
Online exclusive
Ends: 30-Jun-2026
28.40
Save 3.75 (13%)
Each
Chemical Identifiers
CAS3992-42-5
IUPAC Name4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one hydrochloride
Molecular FormulaC9H14ClN3O4
InChI KeyLTKCXZGFJFAPLY-VOBHXVAMNA-N
SMILESCl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only