N-Methyl-D-glucamine, 99%
N-Methyl-D-glucamine, 99%
N-Methyl-D-glucamine, 99%
Thermo Scientific Chemicals

N-Methyl-D-glucamine, 99%

CAS: 6284-40-8 | C7H17NO5 | 195.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number L14282.18
also known as L14282-18
Price (USD)
29.50
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
29.50
Each
Chemical Identifiers
CAS6284-40-8
IUPAC Name6-(methylamino)hexane-1,2,3,4,5-pentol
Molecular FormulaC7H17NO5
InChI KeyMBBZMMPHUWSWHV-UHFFFAOYNA-N
SMILESCNCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Assay (Non-aqueous acid-base Titration)≥98.5% to ≤101.5% (Dry wt. basis)
Water Content (Karl Fischer Titration)≤1.0%
View more
N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methyl-D-glucamine is used in conjunction with iodinated organic compounds in contrast media viz. diatrizoate meglumine and iodipamide meglumine. It is useful in ion-exchange resins to chelate boron ions selectively. It is also used as an excipient in pharmaceuticals, in cosmetics and X-ray contrast media. It is also employed in the synthesis of surface active agents, pharmaceuticals and dyes.

Solubility
Soluble in water.

Notes
Air sensitive and hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Polowczyk, I.; Urbano, B. F.; Rivas, B. L.; Bryjak, M.; Kabay, N. Equilibrium and kinetic study of chromium sorption on resins with quaternary ammonium and N-methyl-d-glucamine groups. Chem. Eng. J. 2016, 284, 395-404.
  2. Samatya, S.; Tuncel, S. A.; Kabay, N. Boron removal from RO permeate of geothermal water by monodisperse poly(vinylbenzyl chloride-co-divinylbenzene) beads containing N-methyl-d-glucamine. Desalination 2015, 364, 75-81.