DL-Valine ethyl ester hydrochloride, 99%
DL-Valine ethyl ester hydrochloride, 99%
DL-Valine ethyl ester hydrochloride, 99%
Thermo Scientific Chemicals

DL-Valine ethyl ester hydrochloride, 99%

CAS: 23358-42-1 | C7H16ClNO2 | 181.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L17906.14
also known as L17906-14
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS23358-42-1
IUPAC Nameethyl 2-amino-3-methylbutanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyPQGVTLQEKCJXKF-UHFFFAOYSA-N
SMILESCl.CCOC(=O)C(N)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
Melting Point (clear melt)101-109?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only