4-Fluoro-L-phenylalanine, 98+%
4-Fluoro-L-phenylalanine, 98+%
4-Fluoro-L-phenylalanine, 98+%
Thermo Scientific Chemicals

4-Fluoro-L-phenylalanine, 98+%

CAS: 1132-68-9 | C9H10FNO2 | 183.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19934.03
also known as L19934-03
Price (USD)
191.65
Special offer
Online exclusive
Ends: 28-Dec-2026
225.00
Save 33.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
191.65
Special offer
Online exclusive
Ends: 28-Dec-2026
225.00
Save 33.35
Each
Chemical Identifiers
CAS1132-68-9
IUPAC Name2-amino-3-(4-fluorophenyl)propanoic acid
Molecular FormulaC9H10FNO2
InChI KeyXWHHYOYVRVGJJY-UHFFFAOYNA-N
SMILESNC(CC1=CC=C(F)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to pale cream
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.0 to ≤103.0% (non-U.S. specification)
Assay from Suppliers CofA≥97.0% (U.S. specification)
Optical Rotation-24° ± 1.5° (c=1, water, non-U.S. specification)
View more
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-L-phenylalanine is a substrate for tyrosine hydroxylase (TH) that has been used to study the regulation of that enzyme.

Solubility
Soluble in water and 0.5M HCl (50 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. R.Shiman; D.W.Gray; M.A.Hill. Regulation of rat liver phenylalanine hydroxylase. I. Kinetic properties of the enzyme's iron and enzyme reduction site. Journal of Biological Chemistry. 1994, 269,(40), 24637-24646.
  2. P.J.Hillas; P.F.Fitzpatrick. A mechanism for hydroxylation by tyrosine hydroxylase based on partitioning of substituted phenylalanines. Biochemistry (Washington). 1996, 35,(22), 6969-6975.