Sambucus Nigra (Elderberry Bark) Lectin (SNA, EBL), fluorescein (FITC)
Invitrogen™

Sambucus Nigra (Elderberry Bark) Lectin (SNA, EBL), fluorescein (FITC)

Sambucus Nigra (Elderberry Bark) Lectin (SNA, EBL), fluorescein (FITC), is a bright-green SNA-fluorophor conjugate. SNA belongs to the group ofRead more
Have Questions?
Catalog NumberQuantity
L324792 mg
Catalog number L32479
Price (USD)
211.56
Special offer
Online exclusive
Ends: 31-Dec-2026
258.00
Save 46.44
Each
Quantity:
2 mg
Price (USD)
211.56
Special offer
Online exclusive
Ends: 31-Dec-2026
258.00
Save 46.44
Each
Sambucus Nigra (Elderberry Bark) Lectin (SNA, EBL), fluorescein (FITC), is a bright-green SNA-fluorophor conjugate. SNA belongs to the group of type 2 ribosome-inactivating proteins and is composed of an A-chain with enzymatic activity and a B-chain with carbohydrate-binding activity. SNA can induce apoptosis through the caspase-dependent pathway at low concentrations. This effect depends on the carbohydrate binding of the B-chain and is independent from the catalytic activity of the A-chain. In colon cancers, it has been reported that 90% of carcinomas express increased levels of sialic acid, linked to galactose residues on the surface of tumor cells, that is detectable with SNA.

View complete list of fluorescent dye-conjugated lectins ›

• Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: sialic acid
• Inhibiting/eluting sugar: 500 mM lactose
• Blood group specificity: non-specific, A > B, O
• Unconjugated lectin molecular weight: 140 KDa
• Immunofluorescence working concentration: 5–20 μg/mL

SNA-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide. 0.1 mM Ca2+.

A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label TypeClassic Dyes
Protein SubtypeLectins
Quantity2 mg
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
SourceElderberry
ConjugateFluorescein
FormSolution
Unit SizeEach
Contents & Storage
When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after reciept.