LIVE/DEAD™ FungaLight™ Yeast Viability Kit, for flow cytometry - FAQs
LIVE/DEAD™ FungaLight™ Yeast Viability Kit, for flow cytometry - FAQs
View additional product information for LIVE/DEAD™ FungaLight™ Yeast Viability Kit, for flow cytometry - FAQs (L34952)
3 product FAQs found
What are the fluorescence excitation/emission maxima for the SYTO 9 dye and propidium iodide in the LIVE/DEAD FungaLight Yeast Viability Kit, for flow cytometry?
SYTO 9 dye: 480/500 nm
Propidium iodide: 490/635 nm
How do I prepare dead cell controls for LIVE/DEAD cell viability assays?
There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.
Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.
Can I look at bacteria and yeast on a flow cytometer?
Yes, you can. We offer:
-Counting assays: Bacteria Counting Kit, for flow cytometry (Cat. No. B7277) or LIVE/DEAD BacLight Bacterial Viability and Counting Kit, for flow cytometry (Cat. No. L34856).
-Viability/vitality assays: LIVE/DEAD BacLight Bacterial Viability Kit (Cat. No. L13152).
-Membrane potential: BacLight Bacterial Membrane Potential Kit (Cat. No. B34950).
-Yeast viability/vitality assays: LIVE/DEAD FungaLight Yeast Viability Kit, for flow cytometry (Cat. No. L34952), FungaLight Yeast CFDA, AM/Propidium Iodide Vitality Kit (Cat. No. F34953)