Low Density Lipoprotein from Human Plasma, Acetylated (AcLDL) - FAQs
Low Density Lipoprotein from Human Plasma, Acetylated (AcLDL) - FAQs
View additional product information for Low Density Lipoprotein from Human Plasma, Acetylated (AcLDL) - FAQs (L35354)
2 product FAQs found
If the Low Density Lipoprotein (LDL) from Human Plasma was accidentally frozen, can I still use it?
We recommend storing Low Density Lipoprotein (LDL) from Human Plasma at 2-8 degrees C. We do not recommend using frozen product.
Can I use Low Density Lipoprotein (LDL) from Human Plasma on other species?
We have not tested this.