Which species of yeast/fungi can I use with the LIVE/DEAD Yeast Viability Kit?
The LIVE/DEAD Yeast Viability Kit has been tested on the following fungi: S. cerevisiae (five different strains), Candida pseudotropicalis, Neurospora crassa and Aspergillus nidulans. A good correlation between the results obtained with the LIVE/DEAD Yeast Viability Kit and those obtained with standard plate counts was achieved with both S. cerevisiae and C. pseudotropicalis. Tests have been performed on both logarithmically growing cultures and cells that have undergone different forms of environmental stress