LIVE/DEAD™ Cell-Mediated Cytotoxicity Kit, for animal cells - FAQs
LIVE/DEAD™ Cell-Mediated Cytotoxicity Kit, for animal cells - FAQs
View additional product information for LIVE/DEAD™ Cell-Mediated Cytotoxicity Kit, for animal cells - FAQs (L7010)
1 product FAQs found
Which optical filter sets are compatible with the LIVE/DEAD Cell-Mediated Cytotoxicity Kit?
The fluorescence from both live and dead bacteria stained with the LIVE/DEAD Cell-Mediated Cytotoxicity Kit may be viewed simultaneously with any standard fluorescein long-pass filter set. Alternatively, the fluorescent live and dead cells may be viewed separately with fluorescein and either rhodamine or Texas Red bandpass filter sets. A summary of the fluorescence microscope filter sets recommended for use with the LIVE/ DEAD Cell-Mediated Cytotoxicity Kit is shown in Table 1 on Page 2 in the manual (https://tools.thermofisher.com/content/sfs/manuals/mp07010.pdf).