What is the fluorescence emission maxima of SYBR 14 dye and propidium iodide?
When bound to DNA, the fluorescence emission maxima of these dyes are 516 nm and 617 nm, respectively. The spectra are available in Figure 1 of the Product Manual.