LysoSensor Blue DND-167 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomesRead more
Have Questions?
Catalog Number
Quantity
L7533
20 x 50 μL
Catalog number L7533
Price (USD)
424.76
Special offer
Online exclusive
Ends: 31-Dec-2026
518.00
Save 93.24
Each
Quantity:
20 x 50 μL
Price (USD)
424.76
Special offer
Online exclusive
Ends: 31-Dec-2026
518.00
Save 93.24
Each
LysoSensor Blue DND-167 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomes more fluorescent in acidic environments. The pKa of LysoSensor Blue DND-167 is ∼5.1.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mM
Detection MethodFluorescence
Emission Wavelength373/425
Quantity20 x 50 μL
Shipping ConditionRoom Temperature
Sub Cellular LocalizationLysosomes
ColorBlue
For Use With (Equipment)Fluorescence Microscope
Product LineLysoSensor
Product TypeDye
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Frequently asked questions (FAQs)
Can I use a plate reader to measure the pH of acidic organelles, such as lysosomes, with LysoSensor ratiometric probes?
Using a plate reader for measuring the pH of acidic organelles, such as lysosomes, with LysoSensor ratiometric probes, provides weak pH measurements, as discrete regions of interest cannot be selected for analysis. Therefore, there is no way of excluding extracellular fluorescence and little to no spatial or morphological information can be obtained. Instead, a readout of whole-cell fluorescence is provided, resulting in the average intraorganellar pH of all labeled organelles for the analyzed cell population. The is also the risk that a signal may be dominated by a few extremely bright, but essentially irrelevant cells, such as dead cells, or dye-labeled debris particles.
For more information about measuring luminal pH in organelles, please see the reference linked below:
What reagents would you recommend for measuring the pH in acidic organelles, such as lysosomes?
For measuring the pH in acidic organelles, such as lysosomes, we recommend our LysoSensor ratio metric probes. Please see the links to the product page and descriptions below:
LysoSensor Yellow/Blue dextran, 10,000 MW, Anionic, Fixable (Cat. No. L22460): LysoSensor Yellow/Blue dextran is a ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes. This dextran conjugate is taken up by the cells through endocytosis. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue dextran is ~3.9.
LysoSensor Yellow/Blue DND-160 - Special Packaging (Cat. No. L7545): LysoSensor Yellow/Blue DND-160 is a ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue DND-160 is ~4.2.
LysoSensor Blue DND-167 - Special Packaging (Cat. No. L7533):
LysoSensor Blue DND-167 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomes more fluorescent in acidic environments. The pKa of LysoSensor Blue DND-167 is ~5.1.
LysoSensor Green DND-189 - Special Packaging (Cat. No. L7535):
LysoSensor Green DND-189 can be used to measure the pH of acidic organelles, such as lysosomes. The LysoSensor dye becomes more fluorescent in acidic environments. The pKa of LysoSensor Green DND-189 is ~5.2.
Expression of the voltage-gated sodium channel NaV1.5 in the macrophage late endosome regulates endosomal acidification.
Authors:Carrithers MD, Dib-Hajj S, Carrithers LM, Tokmoulina G, Pypaert M, Jonas EA, Waxman SG,
Journal:J Immunol
PubMed ID:17548620
'Voltage-gated sodium channels expressed on the plasma membrane activate rapidly in response to changes in membrane potential in cells with excitable membranes such as muscle and neurons. Macrophages also require rapid signaling mechanisms as the first line of defense against invasion by microorganisms. In this study, our goal was to ... More
Fluorescence lifetime characterization of novel low-pH probes.
Authors:Lin HJ, Herman P, Kang JS, Lakowicz JR
Journal:Anal Biochem
PubMed ID:11444806
'The structures and functions of the cellular acidic compartments are strongly dependent on the pH gradients across vesicular membranes. Measurement and imaging of the vesicular pH require fluorophores with appropriate pK(a) values. In this report, we characterized the pH-dependent lifetime responses of a family of acidotropic probes, LysoSensors, to evaluate ... More
Expression and localization of cathepsin k in adult rat sertoli cells.
Authors:Anway MD, Wright WW, Zirkin BR, Korah N, Mort JS, Hermo L
Journal:Biol Reprod
PubMed ID:14585819
'The cathepsins are a family of cysteine proteases that have been broadly implicated in proteolytic processes during cell growth, cell development, and normal adult cellular function. Cathepsin L is a major secretory product of rat and mouse Sertoli cells, the absence of which in furless mice is associated with atrophy ... More
Fluorescent indicators for intracellular pH.
Authors:Han J, Burgess K,
Journal:Chem Rev
PubMed ID:19831417
This review is about intracellular pH sensors, includingsmall fluorescent organic molecules, nanoparticles, andfluorescent proteins, e.g., GFP. It focuses on their preparations, photophysical properties, and advantages/disadvantagesfor intracellular pH measurements. The discussion is limitedto fluorescent indicators that have been applied to measureintracellular pH values since 1980. ... More
Tamoxifen inhibits acidification in cells independent of the estrogen receptor.
Authors:Altan N, Chen Y, Schindler M, Simon SM
Journal:Proc Natl Acad Sci U S A
PubMed ID:10200279
Tamoxifen has been reported to have numerous physiological effects that are independent of the estrogen receptor, including sensitization of resistant tumor cells to many chemotherapeutic agents. Drug-resistant cells sequester weak base chemotherapeutics in acidic organelles away from their sites of action in the cytosol and nucleus. This work reports that ... More