Gel Loading Tips, standard, round - FAQs
Gel Loading Tips, standard, round - FAQs
View additional product information for Gel Loading Tips, standard, round - FAQs (LC1001)
1 product FAQs found
Do you offer gel loading tips to load protein gels?
We offer Invitrogen Gel Loading Tips for easy and accurate loading of samples. They are RNase/DNase-free and non-autoclavable. Three varieties are offered: standard round tips, flat gel tips, and Eppendorf round gel tips.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.