Novex™ Tricine SDS Running Buffer (10X)
Novex™ Tricine SDS Running Buffer (10X)
Novex™ Tricine SDS Running Buffer (10X)
Invitrogen™

Novex™ Tricine SDS Running Buffer (10X)

Novex Tricine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on tricine gels. TricineLeia mais
Have Questions?
Número do catálogoQuantity
LC1675500 mL
Número do catálogo LC1675
Preço (BRL)
1.112,80
Each
Quantity:
500 mL
Preço (BRL)
1.112,80
Each
Novex Tricine SDS Running Buffer (10X) is formulated for separation of proteins in their denatured state on tricine gels. Tricine gels are specifically designed for the resolution of low molecular weight proteins. In this buffer system, tricine substitutes glycine in the running buffer, resulting in more efficient stacking and destacking of low molecular weight proteins and higher resolution of smaller peptides.

Recommended buffers: Use with tricine sample buffer to obtain optimal separation with tricine SDS running buffer.

See all buffers and reagents for SDS-PAGE ›
For Research Use Only. Not for use in diagnostic procedures.
Especificações
Chemical Name or MaterialSample Loading Buffer
Recommended StorageTricine SDS Running buffer (2X) containing 0.1% sodium dodecyl sulfate (SDS), Tris Base, Tricine at pH 8.3

Solubility InformationWater
Concentration10X
Physical FormLiquid
Product LineNovex
Quantity500 mL
Unit SizeEach

Frequently asked questions (FAQs)

Can urea be used with the precast Tricine gel electrophoresis system to achieve denatured results?

Adding urea to the sample and running buffers, in conjunction with SDS, may provide improved solubilization of the sample if denaturation by SDS does not prove to be sufficient. This must be tested empirically for the protein of interest.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.