Product Image
Invitrogen™
Incubation Tray, 10 x 14 cm
The Incubation Tray enables convenient and even staining of E-PAGE™ blots. The tray is 10 cm (l) x 14 cmRead more
Have Questions?
Catalog NumberQuantity
LC21028 each
Catalog number LC2102
Price (USD)
203.65
Online exclusive
212.00
Save 8.35
Each
Quantity:
8 each
Price (USD)
203.65
Online exclusive
212.00
Save 8.35
Each
The Incubation Tray enables convenient and even staining of E-PAGE™ blots. The tray is 10 cm (l) x 14 cm (w) x 3 cm (d) with a lid. It is suitable for use with immunodetection or blot stains.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity8 each
Shipping ConditionRoom Temperature
Product LineNovex
Product TypeIncubation Tray
Unit SizeEach
Contents & Storage
Each pack of Incubation Trays provides 8 plastic trays and 8 lids. Store at room temperature.
Frequently asked questions (FAQs)
If a green color appears on the membrane after the transfer using the iBlot 2 Dry Blotting System, will it affect the results?
The green discoloration is copper deposits from the transfer stack, and it does not affect the results. To minimize this effect, shake excess water off the filter paper and buffer from the gel before placing each on the stack. The current formulation of stacks minimizes the green discoloration.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I cut the bottom stack of my iBlot 2 Transfer Stack to fit my separating gel?
No. Do not trim the iBlot 2 Transfer Stacks to fit your gel size.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is the PVDF membrane in the iBlot 2 PVDF Gel Transfer Stack pre-activated?
The PVDF membrane is pre-activated and ready for use without any pre-treatment with alcohol.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I use the stack and/or absorbent pad more than once when using the iBlot 2 Dry Blotting System?
No. The transfer stacks have a finite amount of ions to drive the transfer and are depleted after a single use.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I perform multiple transfers, with a new transfer starting immediately after the previous one when using the iBlot 2 Dry Blotting System?
Yes. The device is designed to do so with no impact on performance.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.