Novex™ Tris-Glycine SDS Running Buffer (10X), 5 L - FAQs
Novex™ Tris-Glycine SDS Running Buffer (10X), 5 L - FAQs
View additional product information for Novex™ Tris-Glycine SDS Running Buffer (10X) - FAQs (LC2675, LC26754, LC26755)
2 product FAQs found
Where do I find buffer recipes for your precast protein gels?
The formulations of buffers for our precast protein gels can be found at this link: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-gel-electrophoresis/protein-electrophoresis-buffers-reagents.html
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.