Novex™ Tris-Glycine SDS Buffer Kit
Novex™ Tris-Glycine SDS Buffer Kit
Novex™ Tris-Glycine SDS Buffer Kit
Invitrogen™

Novex™ Tris-Glycine SDS Buffer Kit

Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results. All buffers are made with high-purity reagents andRead more
Have Questions?
Catalog NumberQuantity
LC26771 kit
Catalog number LC2677
Price (USD)
52.78
Special offer
Online exclusive
Ends: 27-Jun-2026
56.75
Save 3.97 (7%)
Each
Quantity:
1 kit
Price (USD)
52.78
Special offer
Online exclusive
Ends: 27-Jun-2026
56.75
Save 3.97 (7%)
Each
Pre-mixed buffers are a convenient way to ensure high-quality, consistent electrophoresis results. All buffers are made with high-purity reagents and are strictly quality controlled. The buffers are provided as a concentrated solution requiring a simple dilution with deionized water before use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel TypeSDS PAGE Gel
Quantity1 kit
Gel CompatibilityNovex Tris-Glycine Gel
Product LineNovex
Product TypeBuffer Kit
Unit SizeEach
Contents & Storage
• 500 mL 10X Tris-Glycine SDS Running Buffer
• 20 mL 2X Tris-Glycine SDS Sample Buffer

All Novex™ buffers have a 6 month shelf life. Store at +4°C to 25°C, depending upon formulation.