Novex® IEF Sample Buffer pH 3-10 (2X) is optimized pre-mixed IEF sample buffer for using with Novex® pH 3-10 IEFRead more
Have Questions?
Catalog Number
Quantity
LC5311
25 mL
Catalog number LC5311
Price (USD)
33.80
Each
Quantity:
25 mL
Price (USD)
33.80
Each
Novex® IEF Sample Buffer pH 3-10 (2X) is optimized pre-mixed IEF sample buffer for using with Novex® pH 3-10 IEF gels. IEF is a sensitive technique which is affected by many factors. The optimized pre-mixed IEF buffers reduce variability and ensure consistent results.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferSample Loading Buffers
Concentration2 X
Gel CompatibilityNovex™ IEF Gels
Quantity25 mL
Shipping ConditionWet Ice
Gel TypeIEF
Product LineNovex
Product TypeIEF Sample Buffer
pH Range3 to 10
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).
Frequently asked questions (FAQs)
When staining IEF gels with the Colloidal Blue Staining Kit, is it necessary to use the fixing solution?
Yes, the fix serves two purposes: it fixes the sample in the IEF gel and it helps to remove gel background.
If you do not use the fixing solution, the background on the gels will be high and detection will be less sensitive.
High background is caused by ampholytes remaining in the gel.