SeeBlue™ Plus2 Pre-stained Protein Standard
Invitrogen™

SeeBlue™ Plus2 Pre-stained Protein Standard

SeeBlue Plus2 Pre-Stained Standard contains 10 proteins (4–250 kDa): 8 blue-dyed and 2 with contrasting colors, for easy and quickRead more
Have Questions?
Catalog NumberQuantity
LC5925500 μL
Catalog number LC5925
Price (USD)
240.65
Online Exclusive
256.00
Save 15.35 (6%)
Each
Quantity:
500 μL
Request bulk or custom format
Price (USD)
240.65
Online Exclusive
256.00
Save 15.35 (6%)
Each
SeeBlue Plus2 Pre-Stained Standard contains 10 proteins (4–250 kDa): 8 blue-dyed and 2 with contrasting colors, for easy and quick evaluation of electrophoresis and western transfer efficiency. The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Applications
• Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
• Monitoring protein transfer onto membranes after western blotting
• Sizing of proteins on SDS-PAGE gels and western blots

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodColorimetric
Gel CompatibilityBolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels
Molecular Weight (g/mol)198, 98, 62, 49, 38, 28, 17, 14, 6, 3 kDa
Quantity500 μL
Ready to LoadYes
Shipping ConditionWet Ice
Number of Markers10
Product LineSeeBlue
Product TypeProtein Ladder
Size Range3 to 200 kDa
Stain TypeBlue, Yellow, Pink
System TypeWestern Blotting, SDS-PAGE
Unit SizeEach
Contents & Storage
500 μL (50 applications of 10 μL each) provided in a plastic vial. Loading Buffer consists of Tris-HCl, Formamide, SDS, and Phenol Red.

Store at 4°C.

Frequently asked questions (FAQs)

Can I store the SeeBlue and SeeBlue Plus2 Pre-stained standards in the freezer to increase their shelf-life?

The recommended storage condition for SeeBlue and SeeBlue Plus2 Pre-stained standards is at 4°C. Freeze-thaw cycles, which could result from the standards being shuttled between the bench and the freezer for each use, could degrade the standards over time. If the standards are to be frozen, aliquot them into single use volumes to avoid repeated freeze-thaw.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why are the molecular weight values for the proteins in your prestained standards such as SeeBlue and SeeBlue Plus2 different in different gel types?

The molecular weight values that are stated in conjunction with our standards are given as "apparent" molecular weights. The differences between the apparent molecular weights and the published molecular weights are a result of the covalent attachment of dye to the proteins used in the marker. The bound dye molecules can carry a charge. Of course, this charge changes the ability of the SDS to bind to the protein in addition to contributing directly to the protein's charge. The result is a protein with an altered charge and consequent change in mobility within the gel.

This explains why the proteins in prestained markers run differently from their unstained counterparts. However, it fails to fully explain why there is further difference observed between the same marker in differing gel types (Tris-Glycine vs NuPAGE gels, for example). The reason for this disparity is the different pHs of the gel types. At higher pH values (Tris-Glycine gels), charges are more likely to be protonated. Meanwhile, at the lower, more neutral pH of a NuPAGE gel, the charges are more skewed towards deprotonation, giving the same stained proteins a more negative charge overall. In an SDS PAGE system, more negative charge means more mobility. This is why the same prestained protein will be "larger" on a Tris-Glycine gel than on a NuPAGE gel. In a NuPAGE gel, the lower (approximately neutral) pH causes more overall negative charge, making the apparent molecular weight much lower. This effect generally increases in magnitude with the size of the protein and is greatest with myosin due to the increased number of dye binding sites.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Do you have a molecular weight reference for SeeBlue Pre-stained Protein Standard (Cat. No. LC5625) run on a Tris-Glycine running system?

Unfortunately, we do not have an image reference of the SeeBlue Pre-stained Protein Standard (Cat. No. LC5625) run on a Tris-Glycine running system. However, the image below shows the SeeBlue Plus2 Pre-stained Protein Standard (Cat. No. LC5925) which shares most proteins with the original SeeBlue. All proteins should run according to this table except for phosphorylase, which is not included in the original SeeBlue Pre-Stained Protein Standard.

SeeBlue Plus2 Pre-stained Protein Standard on NuPAGE Novex Bis-Tris 4-12% Gel

Find additional tips, troubleshooting help, and resources within our Protein Standards and Ladders Support Center.

Can I get the concentration of each protein in the SeeBlue Plus2 Pre-stained Protein Standard (Cat. No. LC5925)?

SeeBlue Plus2 Pre-Stained Standard includes 10 recombinant proteins, that resolve in the range of 4 to 250 kDa depending upon the buffer system being used. Please note that this pre-stained protein standard is not designed for quantitation and we do not recommend that you use this standard in the determination of protein concentration.
Estimations of the protein concentration for SeeBlue Plus2 is given below. However, these values are an approximation and should not be used to quantify samples.
Band per 10 µL: Myosin - 2.20 µg; Phosphorylase-b no estimation is available; BSA - 0.75 µg; GDH - 1.25 µg; Alcohol Dehydrogenase - 0.80 µg; Carbonic Anhydrase- 0.90 µg; Myoglobin (blue)* no estimation available; Lysozyme - 2.50 µg; Aprotinin - 1.80 µg and Insulin - 2.50 µg.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

I used one of your pre-stained protein standards for a western transfer and I noticed that the intensity of the band faded from the membrane during the transfer process. Why is this?

The fading is most likely due to detergent in the western blocking/washing solutions that can remove some of the proteins from the membrane. The dye itself will not wash off of the proteins because it is covalently bound. We have found that smaller pore size membranes retain the proteins better during blocking and wash procedures, and hence recommend use of 0.2 µm instead of 0.45 µm membranes for best resolution and protein retention. After transfer, it is a good idea to circle the pre-stained bands with a pencil on the membrane, so band positions can be identified after blocking and processing.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.