Mag-Fura-2, Tetrapotassium Salt, cell impermeant
Invitrogen™

Mag-Fura-2, Tetrapotassium Salt, cell impermeant

Intracellular magnesium indicator that is ratiometric and UV light-excitable.
Have Questions?
Catalog NumberQuantity
M12901 mg
Catalog number M1290
Price (USD)
564.00
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
564.00
Each

Mag-fura-2, tetrapotassium salt is an intracellular magnesium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (I14402).

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Detection MethodFluorescence
Dye TypeMag-Fura-2
Emission Wavelength491 nm
Excitation Wavelength330 nm
Quantity1 mg
Shipping ConditionRoom Temperature
Emission Wavelength Range390 to 405 nm
Excitation Wavelength Range330 to 345 nm
For Use With (Application)Magnesium Indicator
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product TypeMagnesium Indicator
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Citations & References (86)

Citations & References
Abstract
Ca2+ transients in cardiac myocytes measured with high and low affinity Ca2+ indicators.
Authors:Berlin JR,Konishi M
Journal:Biophysical journal
PubMed ID:8274651
Intracellular calcium ion ([Ca2+]i) transients were measured in voltage-clamped rat cardiac myocytes with fura-2 or furaptra to quantitate rapid changes in [Ca2+]i. Patch electrode solutions contained the K+ salt of fura-2 (50 microM) or furaptra (300 microM). With identical experimental conditions, peak amplitude of stimulated [Ca2+]i transients in furaptra-loaded myocytes ... More
Matrix free Mg2+ changes with metabolic state in isolated heart mitochondria.
Authors:Jung DW, Apel L, Brierley GP
Journal:Biochemistry
PubMed ID:2361136
'The concentration of free Mg2+ in the matrix of isolated heart mitochondria has been monitored by using the fluorescent probe furaptra (mag-fura-2). Beef heart mitochondria respiring in a KCl medium in the absence of external Mg2+ maintain free matrix Mg2+ near 0.50 mM. Addition of Pi under these conditions decreases ... More
Modulation of Mg2+ efflux from rat ventricular myocytes studied with the fluorescent indicator furaptra.
Authors:Tursun P, Tashiro M, Konishi M
Journal:Biophys J
PubMed ID:15626700
'The fluorescent Mg(2+) indicator furaptra (mag-fura-2) was introduced into single ventricular myocytes by incubation with its acetoxy-methyl ester form. The ratio of furaptra's fluorescence intensity at 382 and 350 nm was used to estimate the apparent cytoplasmic [Mg(2+)] ([Mg(2+)](i)). In Ca(2+)-free extracellular conditions (0.1 mM EGTA) at 25 degrees C, ... More
Mg2+ buffering in cultured chick ventricular myocytes: quantitation and modulation by Ca2+.
Authors:Koss KL, Putnam RW, Grubbs RD
Journal:Am J Physiol
PubMed ID:8498485
'To characterize the Mg2+ buffering of cultured chick ventricular myocytes, cytosolic Mg2+ was increased by liberating Mg2+ normally chelated by ATP upon total depletion of ATP content. Because the total Mg content and cell volume remained constant during this time, the difference between the amount of Mg2+ liberated (2.7 mM) ... More
Capacitative Ca2+ entry is closely linked to the filling state of internal Ca2+ stores: a study using simultaneous measurements of ICRAC and intraluminal [Ca2+].
Authors:Hofer AM, Fasolato C, Pozzan T
Journal:J Cell Biol
PubMed ID:9442108
'ICRAC (the best characterized Ca2+ current activated by store depletion) was monitored concurrently for the first time with [Ca2+] changes in internal stores. To establish the quantitative and kinetic relationship between these two parameters, we have developed a novel means to clamp [Ca2+] within stores of intact cells at any ... More