Because it is manufactured at our ISO 13485—certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade monobromobimane isRead more
Have Questions?
Catalog Number
Quantity
M20381
25 mg
Catalog number M20381
Price (USD)
300.00
Each
Quantity:
25 mg
Request bulk or custom format
Price (USD)
300.00
Each
Because it is manufactured at our ISO 13485—certified facilities in Eugene, Oregon, we can guarantee that FluoroPure grade monobromobimane is greater than or equal to 98% pure by HPLC.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeMonobromobimane
Quantity25 mg
Shipping ConditionRoom Temperature
Product LineFluoroPure
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Citations & References (235)
Citations & References
Abstract
Determination of plasma homocysteine by high-performance liquid chromatography with fluorescence detection.
Authors:Jacobsen DW, Gatautis VJ, Green R
Journal:Anal Biochem
PubMed ID:2729575
Severe homocystinemia is frequently associated with vascular disease while the pathological consequences of moderate or slightly elevated plasma homocysteine are unknown. Cobalamin and folate deficiencies may result in an elevation of plasma homocysteine. A sensitive and reproducible assay for total plasma homocysteine has been developed. The essential steps in the ... More
Reactivity of mitochondrial sulfhydryl groups toward dithionitrobenzoic acid and bromobimanes under oligomycin-inhibited and uncoupling conditions.
Authors:Freisleben HJ, Fuchs J, Mainka L, Zimmer G
Journal:Arch Biochem Biophys
PubMed ID:2845867
Thiol reactivity was determined in rat heart mitochondria using chromophores of differing polarities: monobromobimane (MB), dithionitrobenzoate (NbS2), and bromobimane-q (MQ). The purpose of this study is to correlate reaction rates of protein thiols in the mitochondrial membrane with the oligomycin-inhibited and uncoupled states: In all cases investigated the reactivity of ... More
Analysis of biological thiols: determination of thiol components of disulfides and thioesters.
Authors:Fenton SS, Fahey RC
Journal:Anal Biochem
PubMed ID:2939769
This report describes a method for using selective cleavage of thioesters to allow differentiation between thioesters and disulfides. The method identifies thiol components (including glutathione, coenzyme A, and cysteine) of low-molecular-weight thioesters and disulfides in cell extracts, as well as thiols bound to protein via thioester or disulfide links. Thioesters ... More
Thiol-specific probes indicate that the beta-chain of platelet glycoprotein Ib is a transmembrane protein with a reactive endofacial sulfhydryl group.
Authors:Kalomiris EL, Coller BS
Journal:Biochemistry
PubMed ID:2934089
We used two membrane-permeable fluorescent reagents, monobromobimane and N-[[5-(dimethylamino)-1-naphthalenyl]sulfonyl]aziridine (N-dansylaziridine), and one membrane-impermeable fluorescent probe, monobromo(trimethylammonio)bimane, all three of which react selectively with protein thiols, to assess the presence of reactive sulfhydryls in the platelet glycoprotein Ib (GPIb) molecule and establish the topology of any GPIb-reactive thiols in the platelet ... More
An electrophoretic profiling method for thiol-rich phytochelatins and metallothioneins.
Authors:Fan TW, Lane AN, Higashi RM
Journal:Phytochem Anal
PubMed ID:15202602
Thiol-rich peptides such as phytochelatins (PCs) and metallothioneins (MTs) are important cellular chelating agents which function in metal detoxification and/or homeostasis. The variations in molecular sizes and lack of chromophores of these peptides make their analysis difficult. This paper reports an electrophoresis-based method for a broad screen of thiol-rich peptides ... More