Human Vascular Smooth Muscle Cell Basal Medium (formerly Medium 231) - FAQs
Human Vascular Smooth Muscle Cell Basal Medium (formerly Medium 231) - FAQs
View additional product information for Human Vascular Smooth Muscle Cell Basal Medium (formerly Medium 231) - FAQs (M231500)
1 product FAQs found
What is the concentration of glucose in Medium 231?
The concentration of glucose in Medium 231 is 4.6 mM.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.