Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL-4 Antibodies

          Invitrogen

          IL-4 Monoclonal Antibody (E10022), Biotin

          View all (114) IL-4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL-4 Monoclonal Antibody (E10022), Biotin

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          IL-4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IL-4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-4 Antibody (M2IL4BI) in WB

          Western blot analysis of IL-4 was performed by loading 1 µg of human recombiannt protein IL-4 (1 µg/lane, Product # RIL4I) in non-reducing sample buffer and 8 µL PageRuler Plus Prestained Protein Ladder (Product # 26619) per well onto a 4-20% Tris-Glycine polyacrylamide gel (Product # WT4202BOX). Proteins were transferred to a nitrocellulose membrane using the G2 Fast Blotter (Product # 62288) and blocked with 5% Milk/TBST for at least 1 hour at room temperature. IL-4 was detected using a biotin conjugated IL-4 mouse monoclonal antibody (P... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-4 Antibody in Western Blot (WB)
          IL-4 Antibody in ELISA (ELISA)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          M2IL4BI

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          ELISA (ELISA)

          0.001-20 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          E10022

          Immunogen

          Recombinant human IL-4 purified from Pichia pastoris

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation

          Form

          Lyophilized

          Concentration

          0.11 mg/mL once reconstituted

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS with 10% trehalose

          Contains

          proprietary preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_417049

          Product Specific Information

          M2IL4BI targets IL-4 in ELISA applications and shows reactivity with Human samples.

          The M2IL4BI immunogen is recombinant human IL-4 purified from Pichia pastoris.

          M2IL4BI detects IL-4 which has a predicted molecular weight of approximately 15 kDa.

          The M2IL4BI IL4 antibody (biotinylated conjugate of clone E10022) has successfully been paired as the detection antibody in a sandwich ELISA with capture antibody M3IL4I (clone E10023). Typical dilutions for sandwich ELISA include 1 µg/mL for coating and 0.125 - 0.25 µg/mL for detection.

          Biotinylated antibody M2IL4BI (clone E10022) and antibody M3IL4I (clone E10023) have successfully been used in combination with recombinant IL4 protein SIL4 in ELISA applications.

          Target Information

          Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: B cell growth factor 1; B_cell stimulatory factor 1; binetrakin; H-IL-4; IL-4v2; IL4; ILN; Interleukin; interleukin 4; Interleukin4; lymphocyte stimulatory factor 1; M-IL-4; MGC79402; pitrakinra

          View more View less

          Gene Aliases: BCGF-1; BCGF1; BSF-1; BSF1; IL-4

          View more View less

          UniProt ID: (Human) P05112

          View more View less

          Entrez Gene ID: (Human) 3565

          View more View less

          Function(s)
          cytokine activity cytokine receptor binding interleukin-4 receptor binding protein binding growth factor activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter immune response JAK-STAT cascade cholesterol metabolic process positive regulation of cell proliferation positive regulation of gene expression negative regulation of epithelial cell migration positive regulation of macroautophagy B cell differentiation positive regulation of cell migration positive regulation of B cell proliferation negative regulation of tumor necrosis factor production positive regulation of interleukin-10 production positive regulation of interleukin-13 production interleukin-4-mediated signaling pathway type 2 immune response positive regulation of T cell proliferation T cell activation macrophage activation positive regulation of tyrosine phosphorylation of STAT protein myeloid dendritic cell differentiation negative regulation of apoptotic process regulation of isotype switching leukocyte activation positive regulation of MHC class II biosynthetic process positive regulation of T cell differentiation negative regulation of osteoclast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of receptor-mediated endocytosis positive regulation of isotype switching to IgE isotypes positive regulation of isotype switching to IgG isotypes negative regulation of inflammatory response regulation of immune response regulation of multicellular organismal process dendritic cell differentiation positive regulation of cold-induced thermogenesis neuroinflammatory response negative regulation of neuroinflammatory response positive regulation of beta-amyloid clearance positive regulation of cellular respiration positive regulation of leukocyte differentiation negative regulation of complement-dependent cytotoxicity negative regulation of cellular response to transforming growth factor beta stimulus negative regulation of endothelial cell apoptotic process positive regulation of T-helper 2 cell cytokine production positive regulation of ATP biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch