Measure-IT™ High-Sensitivity Nitrite Assay Kit
Invitrogen™

Measure-IT™ High-Sensitivity Nitrite Assay Kit

Provides add-and-read convenience and high-throughput scalability to quantitation of nitrite, thiol, and lead/cadmium
Have Questions?
Catalog NumberQuantity
M360511 Kit
Catalog number M36051
Price (USD)
526.00
Each
Quantity:
1 Kit
Price (USD)
526.00
Each

Measure-iT™ assay kits bring add-and-read convenience and high-throughput scalability to the quantitation of nitrite, thiol, and lead/cadmium. The Measure-iT™ High-Sensitivity Nitrite Assay Kit features solid reproducibility over a wide quantitation range, as well as excellent tolerance of common sample contaminants. The assay is linear from 20 to 500 pmoles of nitrite, making it up to 50 times more sensitive than colorimetric assays based on the Griess reagent. The kit includes a nitrite quantitation standard and a protocol that's optimized for microplate-based application.

For Research Use Only. Not for use in diagnostic procedures.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Nitrite Quantitation
Quantity1 Kit
Detection MethodFluorescence
Emission Wavelength Range450 nm
Excitation Wavelength Range365 nm
Product LineMeasure-IT
Product TypeNitrite Assay Kit
Unit SizeEach

Citations & References (4)

Citations & References
Abstract
The intracellular environment of human macrophages that produce nitric oxide promotes growth of mycobacteria.
Authors:Jung JY, Madan-Lala R, Georgieva M, Rengarajan J, Sohaskey CD, Bange FC, Robinson CM,
Journal:
PubMed ID:23774601
Nitric oxide (NO) is a diffusible radical gas produced from the activity of nitric oxide synthase (NOS). NOS activity in murine macrophages has a protective role against mycobacteria through generation of reactive nitrogen intermediates (RNIs). However, the production of NO by human macrophages has remained unclear due to the lack ... More
Comparison of in vitro human endothelial cell response to self-expanding stent deployment in a straight and curved peripheral artery simulator.
Authors:Ghriallais RN, McNamara L, Bruzzi M,
Journal:J R Soc Interface
PubMed ID:23365191
Haemodynamic forces have a synergistic effect on endothelial cell (EC) morphology, proliferation, differentiation and biochemical expression profiles. Alterations to haemodynamic force levels have been observed at curved regions and bifurcations of arteries but also around struts of stented arteries, and are also known to be associated with various vascular pathologies. ... More
A small molecule deubiquitinase inhibitor increases localization of inducible nitric oxide synthase to the macrophage phagosome and enhances bacterial killing.
Authors:Burkholder KM, Perry JW, Wobus CE, Donato NJ, Showalter HD, Kapuria V, O'Riordan MX,
Journal:Infect Immun
PubMed ID:21911458
Macrophages are key mediators of antimicrobial defense and innate immunity. Innate intracellular defense mechanisms can be rapidly regulated at the posttranslational level by the coordinated addition and removal of ubiquitin by ubiquitin ligases and deubiquitinases (DUBs). While ubiquitin ligases have been extensively studied, the contribution of DUBs to macrophage innate ... More
Nitric oxide mediates T cell cytokine production and signal transduction in histidine decarboxylase knockout mice.
Authors:Koncz A, Pasztoi M, Mazan M, Fazakas F, Buzas E, Falus A, Nagy G,
Journal:J Immunol
PubMed ID:17982051
Histamine is a key regulator of the immune system. Several lines of evidence suggest the role of histamine in T cell activation and accelerated Th1 immune response is a hallmark of histidine decarboxylase knockout (HDC-KO) mice, with a complete lack of endogenously produced histamine. According to our previous work, T ... More