Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CaMKII alpha Antibodies

          Invitrogen

          CaMKII alpha Monoclonal Antibody (6G9)

          Advanced Verification
          37 Published Figures
          133 References
          View all (39) CaMKII alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CaMKII alpha Monoclonal Antibody (6G9)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (37)
          • Advanced Verification (2)
          CaMKII alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CaMKII alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 42

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CaMKII alpha Antibody (MA1-048) in WB

          Western blot was performed using Anti-CaMKII alpha Monoclonal Antibody (6G9) (Product # MA1-048) and a band at ~50 kDa corresponding to CaMKII alpha was observed in Mouse and Rat Brain, Mouse and Rat Cerebellum in comparison to other tissues which are reported to be negative. Tissue extracts (30 µg lysate) of Mouse Brain (Lane 1), Rat Brain (Lane 2), Mouse Cerebellum (Lane 3), Rat Cerebellum (Lane 4), Mouse Skeletal Muscle (Lane 5), Rat Skeletal Muscle (Lane 6), Mouse Heart (Lane 7) and Rat Heart (Lane 8) were electrophoresed using Novex® ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Immunocytochemistry (ICC/IF)
          CaMKII alpha Antibody in Immunocytochemistry (ICC/IF)
          CaMKII alpha Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          CaMKII alpha Antibody in Immunocytochemistry (ICC/IF)
          CaMKII alpha Antibody in Immunocytochemistry (ICC/IF)
          CaMKII alpha Antibody in Immunocytochemistry (ICC/IF)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Immunohistochemistry (IHC)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot, Immunoprecipitation (WB, IP)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot, Immunoprecipitation (WB, IP)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody in Western Blot (WB)
          CaMKII alpha Antibody
          CaMKII alpha Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CaMKII alpha Monoclonal Antibody (6G9)

          Product Details

          MA1-048

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-1 µg/mL
          View 55 publications 55 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 48 publications 48 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          -
          View 17 publications 17 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          View 5 publications 5 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Bacteria, Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          6G9

          Immunogen

          Partially purified, rat CaM kinase II.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.17 mg/mL

          Amount

          117 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325403

          Product Specific Information

          MA1-048 detects CaM kinase II from rat and mouse tissues. This antibody has been shown to detect both the phosphorylated and non-phosphorylated alpha subunit of CaM kinase II only. MA1-048 does not detect the ~60 kDa beta subunit in either phosphorylation state.

          MA1-048 has been successfully used in Western blot, immunohistochemistry, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody detects an ~50 kDa protein representing the alpha subunit of CaM kinase II. Immunofluorescence staining of CaM kinase II in rat hippocampal cells with MA1-048 results in intense staining of cell somas and neuropil areas, while nuclei are only lightly stained.

          The MA1-048 immunogen is partially purified, rat CaM kinase II.

          Target Information

          CAMK2A (CaMKII alpha) is a serine/threonine kinase that regulates a number of cellular functions including neuronal processes, gene expression, carbohydrate metabolism, cytoskeletal function, and calcium homeostatsis. Autophosphorylation of threonine 286 in the presence of Ca2+ and calmodulin activates CAMK2A with the ability to switch from calmodulin-dependent to a calmodulin independent activity. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alpha CaM kinase II; alpha-CaMKII; alpha-KAP; Ca2+/calmodulin-dependent protein kinase II alpha; Calci; calcium-calmodulin-dependent protein kinase II; calcium/calmodulin-dependent protein kinase; calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha; calcium/calmodulin-dependent protein kinase II alpha subunit; calcium/calmodulin-dependent protein kinase type II alpha chain; caM kinase II subunit alpha; caM-kinase II alpha chain; CaMK II; caMK-II subunit alpha; EC 2.7.11.17; OTTHUMP00000165787; OTTHUMP00000165788; Phospho-CaMKI; unnamed protein product

          View more View less

          Gene Aliases: CaMKII; mKIAA0968; PK2CDD; PKCCD

          View more View less

          UniProt ID: (Mouse) P11798, (Rat) P11275

          View more View less

          Entrez Gene ID: (Mouse) 12322, (Rat) 25400

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity calmodulin-dependent protein kinase activity protein binding calmodulin binding ATP binding kinase activity GTPase activating protein binding glutamate receptor binding identical protein binding protein homodimerization activity protein serine kinase activity
          Process(es)
          G1/S transition of mitotic cell cycle response to ischemia protein phosphorylation calcium ion transport JAK-STAT cascade positive regulation of cardiac muscle cell apoptotic process peptidyl-serine phosphorylation ionotropic glutamate receptor signaling pathway cellular response to interferon-beta angiotensin-activated signaling pathway positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of JAK-STAT cascade protein autophosphorylation regulation of neurotransmitter secretion regulation of neuronal synaptic plasticity dendrite morphogenesis positive regulation of calcium ion transport dendritic spine development cellular response to interferon-gamma regulation of neurotransmitter receptor localization to postsynaptic specialization membrane neurotransmitter receptor transport to plasma membrane negative regulation of ferroptosis regulation of mitochondrial membrane permeability involved in apoptotic process regulation of protein localization to plasma membrane peptidyl-threonine autophosphorylation regulation of endocannabinoid signaling pathway regulation of neuron migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch