Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Dynein Antibodies

          Invitrogen

          Dynein Monoclonal Antibody (74.1)

          10 References
          View all (5) Dynein antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Dynein Monoclonal Antibody (74.1)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          Dynein Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Dynein Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Dynein Antibody (MA1-070) in ICC/IF

          Immunofluorescent analysis of Dynein in U87-MG Cells. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with a Dynein monoclonal antibody (Product # MA1-070) at a dilution of 1:20 overnight at 4 C, washed with PBS and incubated with a DyLight-488 conjugated secondary antibody (Product # 35503). Dynein staining (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Images were taken at 60X magnification. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Dynein Antibody in Immunocytochemistry (ICC/IF)
          Dynein Antibody in Immunocytochemistry (ICC/IF)
          Dynein Antibody in Immunocytochemistry (ICC/IF)
          Dynein Antibody in Flow Cytometry (Flow)
          Dynein Antibody in Flow Cytometry (Flow)
          Dynein Antibody in Flow Cytometry (Flow)
          Dynein Antibody in Immunoprecipitation (IP)
          Dynein Antibody in RNA Immunoprecipitation (RIP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Dynein Monoclonal Antibody (74.1)

          Product Details

          MA1-070

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          2-4 µg/mL
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          1-2 µg/test
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          View 2 publications 2 publications

          RNA Immunoprecipitation (RIP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Many, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          74.1

          Immunogen

          Purified bovine brain cytoplasmic dynein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2093668

          Product Specific Information

          MA1-070 detects two isoforms of the ~74 kDa polypetide of cytoplasmic dynein.

          MA1-070 has been successfully used in Western blot immunofluorescence, and immunoprecipitation procedures. In Western blot analysis of HeLa cell lysate this antibody detects a ~74 kDa protein representing cytoplasmic dynein.

          The MA1-070 immunogen is purified bovine brain cytoplasmic dynein.

          Target Information

          The directed movement of membrane-bounded organelles is an important process for various cellular functions including membrane transport, secretion, and axonal transport. Cytoplasmic dynein, a ubiquitous minus end-directed microtubule-based motor protein, is believed to be responsible for the retrograde transport of membranous organelles from the synapse to the cell body in fast axonal transport. The cytoplasmic dynein complex is composed of two ~530 kDa heavy chains, and intermediate chains of ~74 kDa and ~53-59 kDa.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 8 kDa dynein light chain; 8 kDa dynein light chain b; 8kD LC; 8kDa LC; cytoplasmic dynein light polypeptide; cytosolic DH IC-1; DLC8b; DNCL1; dynein LC8; dynein light chain-2; dynein, cytoplasmic, light chain 1; dynein, cytoplasmic, light peptide; dynein, cytoplasmic, light polypeptide 1; mPIN; PIN; protein inhibitor of neuronal nitric oxide synthase; protein inhibitor of nitric oxide synthase; radial spoke 22 homolog; unnamed protein product

          View more View less

          Gene Aliases: 1700064A15Rik; 6720463E02Rik; 8kDLC; CDLC2; DLC1; Dlc2; DLC8; DLC8b; DNCL1; DNCL1B; DNCLC1; hdlc1; LC8; LC8a; PIN; RSPH22

          View more View less

          UniProt ID: (Human) P63167, (Bovine) P61285, (Human) Q96FJ2, (Bovine) Q3MHR3, (Mouse) P63168, (Rat) P63170, (Mouse) Q9D0M5, (Rat) Q78P75

          View more View less

          Entrez Gene ID: (Human) 8655, (Bovine) 404151, (Human) 140735, (Bovine) 540369, (Mouse) 56455, (Rat) 58945, (Mouse) 68097, (Rat) 140734

          View more View less

          Function(s)
          enzyme inhibitor activity protein binding enzyme binding protein domain specific binding nitric-oxide synthase regulator activity nitric-oxide synthase inhibitor activity identical protein binding macromolecular complex binding dynein intermediate chain binding scaffold protein binding motor activity deoxyribonuclease inhibitor activity microtubule or microtubule-binding cytoskeletal protein
          Process(es)
          microtubule-based process spermatid development substantia nigra development positive regulation of intracellular transport intraciliary retrograde transport positive regulation of insulin secretion involved in cellular response to glucose stimulus negative regulation of phosphorylation motile cilium assembly positive regulation of mitotic cell cycle spindle assembly checkpoint negative regulation of DNA strand resection involved in replication fork processing apoptotic process viral process cilium movement sperm motility negative regulation of nitric oxide biosynthetic process regulation of mitochondrial membrane potential DNA strand resection involved in replication fork processing mitocytosis positive regulation of nonmotile primary cilium assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch