Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • O-GlcNAc Antibodies

          Invitrogen

          O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (RL2)

          95 Published Figures
          190 References
          View all (11) O-GlcNAc antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite O-linked N-acetylglucosamine (O-GlcNAc) Monoclonal Antibody (RL2)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (95)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 96

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          O-linked N-acetylglucosamine (O-GlcNAc) Antibody (MA1-072) in WB

          Western blot analysis of mouse cortical brain lysates using O-Linked N-Acetylglucosamine Monoclonal Antibody (Product # MA1-072). Blots containing cortical extracts from 4 individual C57BL/6 mice (Lanes 1-4) were blocked with 5% milk in TBST, and probed with MA1-072 (1:1000), followed by a fluorophore-conjugated goat anti-mouse IgG secondary antibody. Data courtesy of the Innovators Program. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunocytochemistry (ICC/IF)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunohistochemistry (IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunohistochemistry (IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunohistochemistry (IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunohistochemistry (IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Immunohistochemistry (IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot, Immunoprecipitation (WB, IP)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot, Immunoprecipitation (WB, IP)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot, Immunoprecipitation (WB, IP)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)
          O-linked N-acetylglucosamine (O-GlcNAc) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA1-072

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 136 publications 136 publications

          Immunohistochemistry (IHC)

          -
          View 17 publications 17 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          View 18 publications 18 publications

          ChIP assay (ChIP)

          Assay-dependent
          View 4 publications 4 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication

          Dot blot (DB)

          1/800
          -

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Chemical

          Published species

          Amphibian, Bovine, Chemical, Fish, Fruit fly, Hamster, Human, Invertebrate, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          RL2

          Immunogen

          Pore complex-lamina fraction purified from rat liver nuclear envelopes.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          200 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_326364

          Product Specific Information

          MA1-072 detects nuclear pore complex (NPC), cytoplasmic and intranuclear O-linked glycoproteins from human, mouse, virus and rat tissues.

          MA1-072 has been successfully used in Western blot, immunofluorescence and immunoprecipitation procedures. By Western blot, this antibody recognizes up to eight different proteins from the NPC of approximately 210, 180, 145, 100, 63, 58, 54 and 45 kDa as well as other O-linked glycoproteins outside of the NPC. Immunofluorescence staining of the NPC with MA1-072 primarily labels NPC O-linked glycoproteins and has been successfully used on a wide variety of mammalian cells. Labeling occurs at the NPC, with most of the labeling at the cytoplasmic and/or nucleoplasmic margins, as well as within the nucleus.

          The MA1-072 immunogen is pore complex-lamina fraction purified from rat liver nuclear envelopes.

          Target Information

          O-linked N-acetylglucosamine (O-GlcNAc) is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine or threonine residues. Unlike other protein glycosylations, O-GlcNAc modifications occur within the nucleus and cytoplasm. They are found on many cellular proteins, including nuclear pore, oncogene, cytoskeletal, heat shock, viral and transcription regulatory proteins. O-GlcNAc glycosylations are thought to obscure phosphorylation sites, counteracting phosphorylation-dependent signaling pathways and protein interactions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: GlcNAc

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch