Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Hepatitis C Virus Core Antigen Antibodies

          Invitrogen

          Hepatitis C Virus Core Antigen Monoclonal Antibody (C7-50)

          72 Published Figures
          163 References
          View all (15) Hepatitis C Virus Core Antigen antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Hepatitis C Virus Core Antigen Monoclonal Antibody (C7-50)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (72)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 72

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Hepatitis C Virus Core Antigen Antibody (MA1-080) in ICC/IF

          Journal of virology 2016 - Published figure using Hepatitis C Virus Core Antigen monoclonal antibody (Product # MA1-080) in Immunocytochemistry {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunocytochemistry (ICC/IF)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry (IHC)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry (IHC)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry (IHC)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry (IHC)
          Hepatitis C Virus Core Antigen Antibody in Immunohistochemistry (IHC)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Western Blot (WB)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Flow Cytometry (Flow)
          Hepatitis C Virus Core Antigen Antibody in Immunoprecipitation (IP)
          Hepatitis C Virus Core Antigen Antibody in ELISA (ELISA)
          Hepatitis C Virus Core Antigen Antibody in Immunomicroscopy (IM)
          Hepatitis C Virus Core Antigen Antibody in Immunomicroscopy (IM)
          Hepatitis C Virus Core Antigen Antibody in In Situ Hybridization (ISH)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA1-080

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 62 publications 62 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 75 publications 75 publications

          Flow Cytometry (Flow)

          1/100
          View 7 publications 7 publications

          ELISA (ELISA)

          Assay-dependent
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 2 publications 2 publications

          Immunomicroscopy (IM)

          -
          View 2 publications 2 publications

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Virus

          Published species

          Virus

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          C7-50

          Immunogen

          Purified HCV core-GST fusion protein (genotype 1b).

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325417

          Product Specific Information

          MA1-080 detects hepatitis C virus (HCV) core protein from transfected human and primate cell lines.

          MA1-080 has been successfully used in Western blot, immunoprecipitation, immunofluorescence and ELISA procedures. By Western blot, this antibody detects a single ~21 kDa protein representing HCV core protein in various transfected cell lines. Immunofluorescence staining of HCV core protein in transfected chimp hepatocytes yields a staining pattern consistent with cytoplasmic and vesicular staining.

          The MA1-080 immunogen is purified HCV core-GST fusion protein (genotype 1b). This antibody recognizes an epitope between amino acid residues 21-40 of HCV core protein. This sequence is conserved among different HCV strains.

          Target Information

          HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HCV core; HepC; HepC core

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch