Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Prolactin Antibodies

          Invitrogen

          Prolactin Monoclonal Antibody (6F11)

          3 Published Figures
          4 References
          View all (71) Prolactin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Prolactin Monoclonal Antibody (6F11)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (3)
          Prolactin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Prolactin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Prolactin Antibody (MA1-10597) in IHC

          Bosnian journal of basic medical sciences 2021 - FIGURE 3 The effect of L-dopa on the expression of PRL and Syt-IV. (A) Densitometric quantification of PRL immunosignal shows that the expression of PRL was significantly higher in treatment groups that received E2 (N = 10) compared to healthy animals (N = 5). Treatment with L-dopa did not have a significant effect on the expression of PRL. (B) Densitometric quantification of Syt-IV immunosignal shows that ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Prolactin Antibody in Immunohistochemistry (IHC)
          Prolactin Antibody in Immunohistochemistry (IHC)
          Prolactin Antibody in Immunohistochemistry (IHC)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Prolactin Monoclonal Antibody (6F11)

          Product Details

          MA1-10597

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-10 µg/mL
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          1-10 µg/mL
          View 2 publications 2 publications

          ELISA (ELISA)

          1-10 µg/mL
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Non-human primate, Sheep, Rat

          Published species

          Human, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          6F11

          Immunogen

          Sheep and human Prolactin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.7 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2284419

          Product Specific Information

          MA1-10597 detects Prolactin from human, mouse, rat, bovine, ovine and squirrel monkey samples.

          MA1-10597 has been successfully used in ELISA, immunohistochemistry, and Western blot applications.

          The MA1-10597 immunogen is sheep and human Prolactin.

          Target Information

          Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: decidual prolactin; growth hormone A1; Human prolactin; luteotropic hormone; luteotropin; precursor to secreted pituitary hormone; prolactin; prolactin family 1, subfamily a, member 1; prolactin peptide; prolactin precursor; prolactin protein; secretory protein; unnamed protein product

          View more View less

          Gene Aliases: GHA1; pPRL; Prl1a1; PRLB; PRLSD1; Prol; RATPRLSD1; RNPROL

          View more View less

          UniProt ID: (Human) P01236, (Sheep) P01240, (Bovine) P01239, (Rat) P01237

          View more View less

          Entrez Gene ID: (Human) 5617, (Sheep) 443317, (Bovine) 280901, (Rat) 24683, (Mouse) 19109

          View more View less

          Function(s)
          prolactin receptor binding hormone activity protein binding peptide hormone
          Process(es)
          negative regulation of endothelial cell proliferation cell surface receptor signaling pathway female pregnancy negative regulation of angiogenesis mammary gland development response to nutrient levels prolactin signaling pathway positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of JAK-STAT cascade positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of lactation blastocyst formation lactation biosynthetic process response to mechanical stimulus positive regulation of gene expression negative regulation of gene expression negative regulation of nitric oxide mediated signal transduction signal transduction involved in regulation of gene expression peptide hormone secretion response to food negative regulation of apoptotic process response to external biotic stimulus positive regulation of nitric oxide biosynthetic process positive regulation of fatty acid biosynthetic process positive regulation of endocytosis long-day photoperiodism positive regulation of NF-kappaB transcription factor activity positive regulation of NIK/NF-kappaB signaling regulation of meiotic cell cycle process involved in oocyte maturation response to L-arginine positive regulation of endothelial cell proliferation parturition response to nutrient circadian rhythm positive regulation of cell proliferation response to xenobiotic stimulus gene expression positive regulation of vascular endothelial growth factor production negative regulation of hydrogen peroxide biosynthetic process regulation of ossification response to estradiol response to lipopolysaccharide cellular response to hormone stimulus multicellular organismal response to stress response to trichostatin A response to macrophage colony-stimulating factor response to cocaine ovulation cycle maternal behavior negative regulation of nitric oxide biosynthetic process response to ethanol regulation of JAK-STAT cascade positive regulation of epithelial cell proliferation cellular response to ethanol cellular response to hypoxia response to alcohol negative regulation of estrogen biosynthetic process negative regulation of progesterone biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0