Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PTGER2/PTGER3 Antibodies

          Invitrogen

          PTGER2/PTGER3 Monoclonal Antibody (3F12)

          View all (2) PTGER2/PTGER3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PTGER2/PTGER3 Monoclonal Antibody (3F12)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          MA1-12677

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3F12

          Immunogen

          Recombinant human EP2 and EP3 receptor protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.11 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS

          Contains

          0.08% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2269160

          Product Specific Information

          A suggested positive control for this product is porcine brain lysate.

          Target Information

          This polyclonal antibody was raised in Rabbit against PTGER3 / EP3. This antibody has been conjugated with Unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human PTGER3 / EP3. BLAST analysis of the peptide immunogen showed no homology with other human proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EP2 receptor; EP3 prostanoid receptor EP3-III isoform; G-protein coupled receptor; G-protein coupled receptor; PTGER3; PGE receptor EP2 subtype; PGE receptor EP3 subtype; PGE receptor, EP2 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP2 subtype; PGE2 receptor EP3 subtype; PGE2 receptor splice variant; isoform EP3-I; prostaglandin E receptor 2 (EP2 subtype); prostaglandin E receptor 2 (subtype EP2); prostaglandin E receptor 2 (subtype EP2), 53kD; prostaglandin E receptor 2 (subtype EP2), 53kDa; prostaglandin E receptor 3 (subtype EP3); prostaglandin E receptor EP2 subtype; prostaglandin E receptor EP3 subtype; prostaglandin E receptor subtype EP2; prostaglandin E2 receptor EP2 subtype-like; prostaglandin E2 receptor type 2; prostaglandin receptor (PGE-2); prostanoid EP2 receptor; prostanoid EP3 receptor; unnamed protein product

          View more View less

          Gene Aliases: COX-2; EP2; EP3; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; EP3e; EP3R; lnc003875; PGE2-R; Pgerep3; Ptger-ep2; Ptgerep2; Ptgerep3; Rep3; rEP3a; rEP3b

          View more View less

          UniProt ID: (Human) P43116, (Human) P43115, (Mouse) Q62053, (Rat) Q62928, (Rat) P34980

          View more View less

          Entrez Gene ID: (Human) 5732, (Human) 5733, (Mouse) 19217, (Rat) 81752, (Mouse) 19218, (Rat) 24929

          View more View less

          Function(s)
          G-protein coupled receptor activity prostaglandin E receptor activity prostaglandin receptor activity protein binding prostanoid receptor activity
          Process(es)
          cAMP biosynthetic process inflammatory response G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration negative regulation of cell proliferation response to nematode regulation of circadian sleep/wake cycle, wakefulness positive regulation of bone mineralization response to lipopolysaccharide response to progesterone negative regulation of interleukin-1 alpha production negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production positive regulation of interleukin-10 production positive regulation of interleukin-6 production response to lipid positive regulation of glial cell apoptotic process positive regulation of ion transmembrane transport regulation of cell proliferation vasodilation regulation of circadian sleep/wake cycle, REM sleep positive regulation of neuron apoptotic process positive regulation of osteoblast differentiation positive regulation of ossification negative regulation of smooth muscle contraction positive regulation of circadian sleep/wake cycle, non-REM sleep cAMP metabolic process micturition maternal process involved in female pregnancy negative regulation of mucus secretion cellular response to prostaglandin E stimulus regulation of nitric oxide metabolic process positive regulation of hippocampal neuron apoptotic process negative regulation of hippocampal neuron apoptotic process surfactant secretion positive regulation of p38MAPK cascade negative regulation of compound eye retinal cell apoptotic process positive regulation of glutamate secretion, neurotransmission negative regulation of microglial cell migration positive regulation of gastric mucosal blood circulation negative regulation of endothelin secretion positive regulation of DNA biosynthetic process fever generation positive regulation of protein phosphorylation desensitization of G-protein coupled receptor protein signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of gene expression negative regulation of norepinephrine secretion regulation of norepinephrine secretion intestine smooth muscle contraction urinary bladder smooth muscle contraction bicarbonate transport positive regulation of fever generation receptor internalization negative regulation of interleukin-8 production positive regulation of urine volume positive regulation of vasoconstriction negative regulation of insulin secretion smooth muscle contraction involved in micturition negative regulation of gastric acid secretion positive regulation of mucus secretion negative regulation of adenylate cyclase-activating G protein-coupled receptor signaling pathway cAMP/PKA signal transduction negative regulation of potassium ion transmembrane transport positive regulation of pri-miRNA transcription from RNA polymerase II promoter negative regulation of calcium ion transmembrane transport negative regulation of gastrin-induced gastric acid secretion cellular response to forskolin positive regulation of inhibitory G-protein coupled receptor phosphorylation negative regulation of circadian sleep/wake cycle, wakefulness positive regulation of myofibroblast contraction positive regulation of colon smooth muscle contraction negative regulation of small intestine smooth muscle contraction positive regulation of calcitonin secretion positive regulation of substance P secretion prostaglandin receptor internalization positive regulation of neutrophil extravasation negative regulation of forebrain neuron differentiation negative regulation of apoptotic process negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress cell death response to estradiol cellular response to lipopolysaccharide cellular response to interleukin-1 response to salt
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline