Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SNAP25 Antibodies

          Invitrogen

          SNAP25 Monoclonal Antibody (B-29-F9)

          View all (53) SNAP25 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SNAP25 Monoclonal Antibody (B-29-F9)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          SNAP25 Monoclonal Antibody (B-29-F9)

          Product Details

          MA1-12711

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          B-29-F9

          Immunogen

          Rat SNAP25 and mouse NS1 Cells.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS

          Contains

          0.08% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1087384

          Product Specific Information

          A suggested positive control for this product is cerebellum tissue lysate.

          Target Information

          SNAP25 is a t-SNARE involved in the molecular regulation of neurotransmitter release. It may play a role in the synaptic function of specific neuronal systems. SNAP25 associates with proteins involved in vesicle docking and membrane fusion. It also regulates plasma membrane recycling through its interaction with CENPF. Mutations in the gene can results in myasthenic syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALMS1; blind drunk; CG17676; CG17884; CG40452; Dmel CG40452; Dmel_CG40452; FLJ23079; HGNC:11132; HUMSNAP25B(F); resistance to inhibitors of cholinesterase 4 homolog; SNAP 25; SNAP-25; snp25; SUP; super protein; synaptosomal-associated 25 kDa protein; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25kDa; unnamed protein product

          View more View less

          Gene Aliases: bA416N4.2; Bdr; CMS18; DEE117; dJ1068F16.2; GENA70; RIC-4; RIC4; SEC9; SNAP; SNAP-25; SNAP-25a; SNAP-25B; sp; SUP

          View more View less

          UniProt ID: (Chicken) P60878, (Human) P60880, (Bovine) Q17QQ3, (Mouse) P60879, (Rat) P60881

          View more View less

          Entrez Gene ID: (Chicken) 396444, (Human) 6616, (Bovine) 540853, (Mouse) 20614, (Rat) 25012

          View more View less

          Function(s)
          voltage-gated potassium channel activity SNAP receptor activity protein binding syntaxin-1 binding syntaxin binding SNARE binding lipid binding calcium-dependent protein binding myosin binding protein domain specific binding ion channel binding SNARE protein
          Process(es)
          exocytosis vesicle fusion locomotory behavior associative learning regulation of neuron projection development synaptic vesicle priming synaptic vesicle fusion to presynaptic active zone membrane long-term synaptic potentiation regulation of establishment of protein localization potassium ion transmembrane transport exocytic insertion of neurotransmitter receptor to postsynaptic membrane neurotransmitter receptor internalization neurotransmitter uptake chemical synaptic transmission neurotransmitter secretion synaptic vesicle exocytosis regulation of insulin secretion presynaptic dense core vesicle exocytosis long-term memory endosomal transport insulin secretion positive regulation of vesicle fusion positive regulation of synaptic plasticity SNARE complex assembly positive regulation of hormone secretion calcium ion-regulated exocytosis of neurotransmitter positive regulation of synaptic transmission membrane fusion clathrin-mediated endocytosis regulation of synaptic vesicle cycle positive regulation of voltage-gated calcium channel activity positive regulation of glutamate secretion, neurotransmission short-term synaptic potentiation axonogenesis neuron differentiation positive regulation of insulin secretion regulation of synapse assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline