Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RhoA Antibodies

          Invitrogen

          RhoA Monoclonal Antibody (1B8-1C7)

          Advanced Verification
          3 Published Figures
          6 References
          View all (34) RhoA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RhoA Monoclonal Antibody (1B8-1C7)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (3)
          • Advanced Verification (1)
          RhoA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RhoA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RhoA Antibody (MA1-134) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of Mouse Kidney (Lane1), HeLa (Lane2), NIH/3T3 (Lane3), A-431 (lane 4), PC-12 (lane 5), Jurkat (lane 6), and HEK-293 (lane 7). The blots were probed with Anti-RhoA Mouse Monoclonal Antibody (Product # MA1-134, 1:500-1:2000 µg/mL) and detected by chemiluminescence using Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate (Product # 62-6520, 1:4000 dilution). A 21 kDa band corresponding to RhoA was observed across cell lines tested. Known quantity of protein sa... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RhoA Antibody in Western Blot (WB)
          RhoA Antibody in Western Blot (WB)
          RhoA Antibody in Western Blot (WB)
          RhoA Antibody in Western Blot (WB)
          RhoA Antibody in Western Blot (WB)
          RhoA Antibody in Western Blot, Immunoprecipitation (WB, IP)
          RhoA Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          RhoA Monoclonal Antibody (1B8-1C7)

          Product Details

          MA1-134

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Dog, Human, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1B8-1C7

          Immunogen

          Full-length recombinant His-tagged human RhoA
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2536840

          Product Specific Information

          By Western blot, MA1-134 detects recombinant RhoA, but not recombinant RhoB or recombinant RhoC.

          By Western blot on mouse and rat kidney whole tissue lysates, MA1-134 detects a band of ~22kD corresponding to the expected molecular weight of RhoA. A nonspecific band at ~55kD was also detected in both kidney lysates.

          Target Information

          RhoA regulates a signal transduction pathway linking plasma membrane receptors to the assembly of focal adhesions and actin stress fibers. It is involved in a microtubule-dependent signal that is required for the myosin contractile ring formation during cell cycle cytokinesis, and plays an essential role in cleavage furrow formation. RhoA is required for the apical junction formation of keratinocyte cell-cell adhesion, and serves as a target for the yopT cysteine peptidase from Yersinia pestis, vector of the plague, and Yersinia pseudotuberculosis, which causes gastrointestinal disorders. RhoA stimulates PKN2 kinase activity and may be an activator of PLCE1. It is activated by ARHGEF2, which promotes the exchange of GDP for GTP. It is essential for the SPATA13-mediated regulation of cell migration and adhesion assembly and disassembly. The MEMO1-RHOA-DIAPH1 signaling pathway plays an important role in ERBB2-dependent stabilization of microtubules at the cell cortex. It controls the localization of APC and CLASP2 to the cell membrane, via the regulation of GSK3B activity. In turn, membrane-bound APC allows the localization of the MACF1 to the cell membrane, which is required for microtubule capture and stabilization.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Aplysia ras-related homolog 12; aplysia ras-related homolog A; epididymis secretory sperm binding protein; MGC72339; oncogene RHO H12; plysia ras-related homolog A2; ras homolog gene family, member A; Rho; RhoA; small GTP binding protein RhoA; unnamed protein product

          View more View less

          Gene Aliases: ARH12; ARHA; Arha1; Arha2; EDFAOB; RHO12; RHOH12

          View more View less

          UniProt ID: (Human) P61586, (Rat) P61589, (Mouse) Q9QUI0

          View more View less

          Entrez Gene ID: (Human) 387, (Rat) 117273, (Mouse) 11848

          View more View less

          Function(s)
          GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding myosin binding protein kinase binding GDP binding protein domain specific binding Rho GDP-dissociation inhibitor binding
          Process(es)
          mitotic cytokinesis alpha-beta T cell lineage commitment aortic valve formation small GTPase mediated signal transduction Rho protein signal transduction negative regulation of cell-substrate adhesion cell migration substantia nigra development actin cytoskeleton organization regulation of cell migration positive regulation of cell migration cytoplasmic microtubule organization positive regulation of cytokinesis regulation of actin cytoskeleton organization regulation of osteoblast proliferation cell junction assembly substrate adhesion-dependent cell spreading Roundabout signaling pathway cleavage furrow formation apolipoprotein A-I-mediated signaling pathway odontogenesis positive regulation of I-kappaB kinase/NF-kappaB signaling stress fiber assembly apical junction assembly beta selection endothelial cell migration ossification involved in bone maturation wound healing, spreading of cells establishment of epithelial cell apical/basal polarity positive regulation of neuron differentiation negative regulation of cell size positive regulation of alpha-beta T cell differentiation negative chemotaxis positive regulation of stress fiber assembly regulation of focal adhesion assembly Wnt signaling pathway, planar cell polarity pathway positive regulation of lipase activity bone trabecula morphogenesis regulation of microtubule cytoskeleton organization cellular response to lipopolysaccharide cellular response to cytokine stimulus semaphorin-plexin signaling pathway positive regulation of protein serine/threonine kinase activity negative regulation of cell migration involved in sprouting angiogenesis mitotic spindle assembly negative regulation of oxidative phosphorylation endothelial tube lumen extension regulation of modification of postsynaptic structure positive regulation of NIK/NF-kappaB signaling skeletal muscle satellite cell migration negative regulation of reactive oxygen species biosynthetic process mitotic cleavage furrow formation positive regulation of leukocyte adhesion to vascular endothelial cell regulation of modification of postsynaptic actin cytoskeleton cellular response to chemokine positive regulation of T cell migration meiotic spindle organization cell morphogenesis response to hypoxia kidney development angiotensin mediated vasoconstriction involved in regulation of systemic arterial blood pressure regulation of systemic arterial blood pressure by endothelin regulation of transcription from RNA polymerase II promoter cytoskeleton organization microtubule depolymerization cell adhesion cell-matrix adhesion skeletal muscle tissue development regulation of actin polymerization or depolymerization response to xenobiotic stimulus response to mechanical stimulus response to glucose regulation of neuron projection development negative regulation of neuron projection development cerebral cortex cell migration forebrain radial glial cell differentiation cell differentiation positive regulation of cell growth androgen receptor signaling pathway positive regulation of actin filament polymerization establishment or maintenance of actin cytoskeleton polarity stress-activated protein kinase signaling cascade negative regulation of intracellular steroid hormone receptor signaling pathway embryonic cleavage polar body extrusion after meiotic divisions negative regulation of I-kappaB kinase/NF-kappaB signaling response to amino acid negative regulation of neuron apoptotic process positive regulation of neuron apoptotic process response to ethanol negative regulation of neuron differentiation positive regulation of translation positive regulation of cell adhesion positive regulation of vasoconstriction positive regulation of smooth muscle contraction GTP metabolic process neuron projection morphogenesis regulation of dendrite development response to glucocorticoid spindle localization regulation of calcium ion transport negative regulation of synapse assembly cellular response to progesterone stimulus positive regulation of podosome assembly positive regulation of vascular smooth muscle contraction negative regulation of vascular smooth muscle cell proliferation negative regulation of vascular associated smooth muscle cell migration regulation of neural precursor cell proliferation integrin-mediated signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline