LAMP1 Monoclonal Antibody (LY1C6) - FAQs
LAMP1 Monoclonal Antibody (LY1C6) - FAQs
View additional product information for LAMP1 Monoclonal Antibody (LY1C6) - FAQs (MA1164)
1 product FAQs found
Do you recommend aliquoting the LAMP1 Monoclonal Antibody (LY1C6) (Cat. No. MA1-164)?
Yes, we recommend aliquoting the antibody into smaller volumes and then storing the aliquots at -20 degrees C.