Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • p300 Antibodies

          Invitrogen

          p300 Monoclonal Antibody (RW105)

          Advanced Verification
          1 Published Figure
          4 References
          View all (28) p300 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite p300 Monoclonal Antibody (RW105)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          • Published Figures (1)
          • Advanced Verification (2)
          p300 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          p300 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          p300 Antibody (MA1-16622) in ICC/IF

          Immunofluorescence analysis of p300 Monoclonal Antibody (RW105) was performed using 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with p300 Monoclonal Antibody (RW105) (Product # MA1-16622) at 1:100 dilution in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Ale... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          p300 Antibody in Immunocytochemistry (ICC/IF)
          p300 Antibody in Immunocytochemistry (ICC/IF)
          p300 Antibody in Immunocytochemistry (ICC/IF)
          p300 Antibody in Immunocytochemistry (ICC/IF)
          p300 Antibody in Immunocytochemistry (ICC/IF)
          p300 Antibody in Western Blot (WB)
          p300 Antibody in Western Blot (WB)
          p300 Antibody in Flow Cytometry (Flow)
          p300 Antibody in Flow Cytometry (Flow)
          p300 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          p300 Antibody
          p300 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          p300 Monoclonal Antibody (RW105)

          Product Details

          MA1-16622

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:250-1:500
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          1 µg/mL
          -

          Immunoprecipitation (IP)

          1:10-1:500
          -

          ChIP assay (ChIP)

          Assay-Dependent
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mustelid, Mouse, Non-human primate, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          RW105

          Immunogen

          Fusion protein containing residues 1572-2371 of human KAT3B/p300.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_568668

          Product Specific Information

          Suggested positive control: HeLa nuclear extract.

          Target Information

          EP300 encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. p300 is related by sequence to CPB (CREB-binding protein [CREB: cyclic-AMP responsive element binding protein]), and like CPB can stimulate transcription through activation of CREB. This EP300 activity is specifically inhibited by the adenovirus oncoprotein E1A. EP300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; histone butyryltransferase p300; histone crotonyltransferase p300; p300 HAT; protein 2-hydroxyisobutyryltransferase p300; protein isonicotinyltransferase p300; protein lactyltransferas p300; protein propionyltransferase p300; unnamed protein product

          View more View less

          Gene Aliases: A430090G16; A730011L11; KAT3B; MKHK2; p300; p300 HAT; RSTS2

          View more View less

          UniProt ID: (Human) Q09472, (Mouse) B2RWS6

          View more View less

          Entrez Gene ID: (Human) 2033, (Mouse) 328572, (Rat) 170915

          View more View less

          Function(s)
          transcription cofactor binding transcription coactivator binding p53 binding DNA binding chromatin binding damaged DNA binding transcription cofactor activity transcription coactivator activity histone acetyltransferase activity lysine N-acetyltransferase activity, acting on acetyl phosphate as donor protein binding beta-catenin binding zinc ion binding H3 histone acetyltransferase activity H4 histone acetyltransferase activity acetyltransferase activity transferase activity, transferring acyl groups ligand-dependent nuclear receptor binding chromatin DNA binding histone acetyltransferase activity (H3-K18 specific) histone H2B acetyltransferase activity histone acetyltransferase activity (H3-K27 specific) tau protein binding androgen receptor binding NF-kappaB binding RNA polymerase II sequence-specific DNA binding transcription factor binding peptide-lysine-N-acetyltransferase activity protein propionyltransferase activity pre-mRNA intronic binding STAT family protein binding peptide 2-hydroxyisobutyryltransferase activity peptide lactyltransferase (CoA-dependent) activity histone lactyltransferase (CoA-dependent) activity acetylation-dependent protein binding peptide crotonyltransferase activity peptide butyryltransferase activity histone crotonyltransferase activity histone butyryltransferase activity histone isonicotinyltransferase activity DNA-binding transcription factor binding histone H3K122 acetyltransferase activity histone H1K75 acetyltransferase activity RNA polymerase II core promoter proximal region sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding antigen binding protein kinase binding glucocorticoid receptor binding peroxisome proliferator activated receptor binding bHLH transcription factor binding macromolecular complex binding SMAD binding mitogen-activated protein kinase binding histone reader activity protein antigen binding promoter-specific chromatin binding
          Process(es)
          autophagosome assembly negative regulation of transcription from RNA polymerase II promoter response to hypoxia somitogenesis positive regulation of protein phosphorylation thigmotaxis behavioral defense response gluconeogenesis glycolytic process regulation of glycolytic process chromatin remodeling regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter protein acetylation internal protein amino acid acetylation apoptotic process I-kappaB kinase/NF-kappaB signaling JAK-STAT cascade heart development skeletal muscle tissue development learning or memory circadian rhythm lipid biosynthetic process response to xenobiotic stimulus response to glucose organ morphogenesis negative regulation of autophagy positive regulation of glycoprotein biosynthetic process positive regulation of gene expression regulation of mitochondrion organization positive regulation of neuron projection development positive regulation of muscle atrophy N-terminal peptidyl-lysine acetylation internal peptidyl-lysine acetylation peptidyl-lysine acetylation B cell differentiation platelet formation positive regulation of cell growth lung development positive regulation of transforming growth factor beta receptor signaling pathway negative regulation of protein complex assembly protein destabilization cellular response to nutrient levels negative regulation of protein oligomerization positive regulation of collagen biosynthetic process cellular response to UV multicellular organism growth megakaryocyte development endodermal cell differentiation swimming TORC1 signaling TORC2 signaling positive regulation of protein import into nucleus intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator protein kinase B signaling response to estrogen positive regulation by host of viral transcription fat cell differentiation negative regulation of gluconeogenesis positive regulation of translation positive regulation of axon extension positive regulation of cell size positive regulation of gene expression, epigenetic positive regulation of proteolysis positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JAK-STAT cascade positive regulation of protein secretion protein stabilization cartilage development response to calcium ion positive regulation of protein kinase B signaling canonical Wnt signaling pathway regulation of angiotensin metabolic process positive regulation of sarcomere organization face morphogenesis regulation of androgen receptor signaling pathway positive regulation of cell growth involved in cardiac muscle cell development peptidyl-lysine propionylation protein-DNA complex assembly protein localization to chromatin cellular response to leucine cellular response to interleukin-1 response to dexamethasone tricarboxylic acid metabolic process regulation of tubulin deacetylation peptidyl-lysine crotonylation peptidyl-lysine butyrylation DNA repair-dependent chromatin remodeling positive regulation of cellular response to hypoxia positive regulation of hydrogen peroxide-mediated programmed cell death positive regulation of protein acetylation negative regulation of chromosome condensation positive regulation of TORC1 signaling positive regulation of TORC2 signaling positive regulation of T-helper 17 cell lineage commitment negative regulation of miRNA metabolic process regulation of nucleotide-excision repair negative regulation of apoptotic signaling pathway memory response to cobalt ion response to retinoic acid response to tumor necrosis factor cellular response to trichostatin A response to hydrogen peroxide response to ethanol digestive tract development cellular response to hydrogen peroxide response to fatty acid cellular response to antibiotic cellular response to retinoic acid cellular response to cAMP cellular response to glucose stimulus cellular response to mineralocorticoid stimulus cellular response to xenobiotic stimulus cellular response to dexamethasone stimulus cellular response to curcumin cellular response to nerve growth factor stimulus stimulatory C-type lectin receptor signaling pathway nervous system development regulation of autophagy regulation of cellular response to heat positive regulation of protein localization to nucleus regulation of signal transduction by p53 class mediator negative regulation of brown fat cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-7d6nq:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline