Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cardiac Myosin Heavy Chain Antibodies

          Invitrogen

          Cardiac Myosin Heavy Chain Monoclonal Antibody (3-48)

          Advanced Verification
          1 Published Figure
          10 References
          View all (2) Cardiac Myosin Heavy Chain antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cardiac Myosin Heavy Chain Monoclonal Antibody (3-48)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          • Advanced Verification (1)
          Cardiac Myosin Heavy Chain Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Cardiac Myosin Heavy Chain Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cardiac Myosin Heavy Chain Antibody (MA1-26180) in WB

          Western blot was performed using Anti-Cardiac Myosin Heavy Chain Monoclonal Antibody (3-48) (Product # MA1-26180) and a ~ 223 kDa band corresponding to Cardiac Myosin Heavy Chain was observed in Mouse heart, Mouse skeletal muscle, Rat heart, Rat skeletal muscle and was not observed in Mouse liver, Mouse brain, Mouse kidney, Mouse lung, Rat liver and Rat brain. Tissue extracts (30 µg lysate) of Mouse Heart (... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cardiac Myosin Heavy Chain Antibody in Western Blot (WB)
          Cardiac Myosin Heavy Chain Antibody in Western Blot (WB)
          Cardiac Myosin Heavy Chain Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Cardiac Myosin Heavy Chain Monoclonal Antibody (3-48)

          Product Details

          MA1-26180

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 3 publications 3 publications

          ELISA (ELISA)

          Assay-dependent
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat

          Published species

          Human, Mouse, Pig, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3-48

          Immunogen

          Human ventricular myosin.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2147173

          Product Specific Information

          This antibody is myosin alpha and beta heavy chain specific. Affinity constants: Human ventricular myosin heavy chains - 3.33 x 108M-1; Human atrial myosin heavy chains - 1.48 x 108M-1; Human skeletal muscle myosin - 1.06 x 108M-1; Canine cardiac myosin heavy chains - 2.06 x 108M-1. This antibody cross-reacts with heavy chains of human myosin isolated from slow skeletal muscle, fast skeletal muscle, eye muscle, and streptococcal M5 peptide. It does not react with human platelets, smooth muscle myosin heavy chains, myosin light chains, myosin fragments circulating in blood of patients with myocardial infarction, nor does it react with any other human tissue antigens or blood cells.

          Recommended positive control: Slow skeletal muscle, fast skeletal muscle, eye muscle.

          Target Information

          Cardiac Myosin Heavy Chain (MHC) is a crucial component of the contractile machinery in heart muscle cells. It primarily forms part of the thick filament in sarcomeres, the fundamental units of muscle contraction. The cardiac myosin heavy chain comes in two isoforms, alpha-MHC and beta-MHC, which are encoded by the MYH6 and MYH7 genes, respectively. The alpha-MHC isoform is predominant in the atria and is characterized by faster ATPase activity and contraction speed, whereas the beta-MHC isoform is predominant in the ventricles and is associated with greater endurance. Each myosin molecule consists of two heavy chains, two essential light chains (ELC), and two regulatory light chains (RLC). The heavy chains have distinct functional regions: the globular head, which binds to actin and hydrolyzes ATP to produce force, and the helical tail, which organizes into the thick filament backbone. Mutations in cardiac MHC can lead to various cardiomyopathies, including hypertrophic cardiomyopathy (HCM) and dilated cardiomyopathy (DCM), both of which significantly affect the heart's functionality. These disorders underscore the importance of MHC in maintaining normal cardiac physiology and highlight its complex regulation and mechanotransduction capabilities.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alpha cardiac MHC; alpha myosin; alpha-myosin heavy chain; antigen MLAA-21; beta myosin heavy chain; beta-myosin heavy chain; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain 21/26; cardiac myosin heavy chain 5; cardiomyopathy, hypertrophic 1; muscle-type myosin II; myHC-alpha; myHC-beta; myHC-slow; myosin 7; myosin heavy chain; myosin heavy chain 6; Myosin heavy chain 7; myosin heavy chain beta-subunit; myosin heavy chain MYH7; muscle filament; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain polypeptide 7 cardiac muscle fetal; myosin heavy chain slow isoform; myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, adult; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, polypeptide 7; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy chain 7B, cardiac muscle, beta; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy polypeptide 7, cardiac muscle, beta; myosin, heavy polypeptide 7B, cardiac muscle, beta; rhabdomyosarcoma antigen MU-RMS-40.7A; sarcomeric protein; slow A MYH14; type slow/beta myosin heavy chain; U937-associated antigen; unnamed protein product

          View more View less

          Gene Aliases: A830009F23Rik; alpha-MHC; alphaMHC; ASD3; B-MHC; beta-MHC; betaMHC; Bmyo; CMD1EE; CMD1S; CMH1; CMH14; CMYO7A; CMYO7B; CMYP7A; CMYP7B; lncMYH7b; MHC14; MPD1; MYH-beta/slow; MYH14; Myh7; MYHC; Myhc-a; Myhc-b; MyHC-beta; MyHC-I; MyHC-slow; MYHCA; MYHCB; Myosin-7; SPMD; SPMM; SSS3

          View more View less

          UniProt ID: (Human) P13533, (Human) P12883, (Bovine) Q9BE39, (Human) A7E2Y1, (Mouse) Q02566, (Rat) P02564, (Mouse) Q91Z83, (Mouse) A2AQP0

          View more View less

          Entrez Gene ID: (Human) 4624, (Human) 4625, (Bovine) 282714, (Human) 57644, (Mouse) 17888, (Rat) 29556, (Rat) 29557, (Mouse) 140781, (Mouse) 668940, (Rat) 311570

          View more View less

          Function(s)
          microfilament motor activity motor activity ATP binding myosin phosphatase activity protein kinase binding actin filament binding protein binding calmodulin binding calcium-dependent ATPase activity identical protein binding macromolecular complex binding ATPase activity
          Process(es)
          regulation of the force of heart contraction regulation of heart rate skeletal muscle contraction muscle contraction striated muscle contraction adult heart development regulation of the force of skeletal muscle contraction muscle filament sliding regulation of slow-twitch skeletal muscle fiber contraction ATP metabolic process ventricular cardiac muscle tissue morphogenesis cardiac muscle contraction muscle system process transition between fast and slow fiber cardiac muscle hypertrophy in response to stress cellular response to hydrogen peroxide cellular response to angiotensin cellular response to 3,3',5-triiodo-L-thyronine in utero embryonic development visceral muscle development regulation of heart contraction regulation of blood pressure actin filament-based movement myofibril assembly regulation of ATPase activity sarcomere organization atrial cardiac muscle tissue morphogenesis cardiac muscle cell development regulation of heart growth regulation of calcium-mediated signaling regulation of cytosolic calcium ion concentration regulation of cardiac muscle cell contraction regulation of CAMKK-AMPK signaling cascade
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch