Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • P-Glycoprotein (ABCB1) Antibodies

          Invitrogen

          P-Glycoprotein Monoclonal Antibody (C219)

          Advanced Verification
          19 Published Figures
          49 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite P-Glycoprotein Monoclonal Antibody (C219)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (19)
          • Advanced Verification (2)
          P-Glycoprotein Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          P-Glycoprotein Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          P-Glycoprotein Antibody (MA1-26528) in WB

          Western blot was performed using Anti-P-Glycoprotein Monoclonal Antibody (C219) (Product # MA1-26528) and due to glycosylation, 160-230 kDa band corresponding to ABCB1 was observed across cell lines tested except MCF-7 which is reported to be negative. Whole cell extracts (30 µg lysate) of Hep G2 (Lane 1), Caco-2 (Lane 2), MDCK (Lane 3) and MCF-7 (Lane 4) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # LC2001) by XCell Sur... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Immunocytochemistry (ICC/IF)
          P-Glycoprotein Antibody in Immunohistochemistry (IHC)
          P-Glycoprotein Antibody in Immunohistochemistry (IHC)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Western Blot (WB)
          P-Glycoprotein Antibody in Flow Cytometry (Flow)
          P-Glycoprotein Antibody
          P-Glycoprotein Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          P-Glycoprotein Monoclonal Antibody (C219)

          Product Details

          MA1-26528

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:50-1:500
          View 32 publications 32 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          ≥1:25
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          Assay-dependent
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Fish, Human, Mouse, Rat, Zebrafish

          Published species

          Dog, Fish, Human, Insect, Mouse, Rat, Yeast

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          C219

          Immunogen

          SDS-solubilized plasma membranes of a multidrug resistant Chinese hamster ovary (CHO) cell line and a human cell line.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Purification

          purified

          Storage buffer

          PBS with 1% BSA

          Contains

          0.1% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_795165

          Product Specific Information

          Recommended positive controls: Cell lines or frozen tissue, such as liver or colon.

          MA1-26528 is expected to cross react with a wide range of mammals due to sequence homology.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          This clone recognizes an internal, highly conserved amino acid sequence (VQEALD and VQAALD) found in both protein isoforms, MDR1 P-glycoprotein (~170kDa) and MDR3 P-glycoprotein (~140kDa). In Western blot, boiling samples may reduce visibility of the protein.

          For flow cytometry, cells should be fixed in 3.7% formaldehyde for 10 min at room temperature to permeabilize the cell membrane. Incubate cells with 5-10 µg antibody for 30-60 min at 4°C.

          Target Information

          The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ABC superfamily; P-glycoprotein; ATP binding cassette subfamily B member 1; ATP-binding cassette sub-family B (MDR/TAP) member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette sub-family B member 1; ATP-binding cassette transport protein; ATP-binding cassette, sub-family B (MDR/TAP), member 1; ATP-binding cassette, sub-family B (MDR/TAP), member 1 (P-glycoprotein/multidrug resistance 1); ATP-binding cassette, sub-family B (MDR/TAP), member 1A; ATP-binding cassette, sub-family B (MDR/TAP), member 1B; ATP-binding cassette, sub-family B (MDR/TAP), member 4; ATP-binding cassette, subfamily B (MDR/TAP), member 1A; ATP-binding cassette, subfamily B (MDR/TAP), member 1B; ATP-binding cassette, subfamily B, member 1A; ATP-binding cassette, subfamily B, member 1B; colchicin sensitivity; doxorubicin resistance; ecotropic viral integration site 32; MDR1; P-glycoprotein; mdr1a protein; multi-drug resistance 3; multidrug resistance p-glycoprotein; multidrug resistance protein 1; multidrug resistance protein 1B; multidrug resistance protein 3; multidrug-resistance; multiple drug resistant 1a; P glycoprotein 1; P-glycoprotein; P-glycoprotein 1; P-glycoprotein 2/ multidrug resistance 1b; P-glycoprotein 3; P-glycoprotein/multidrug resistance 1; P-glycoprotein; mdr1a; phospholipid transporter ABCB1; transporter; ABCB1b; unnamed protein product; uORF

          View more View less

          Gene Aliases: ABC20; Abcb1; Abcb4; CD243; CLCS; ENPAT; Evi32; GP170; mdr; mdr-3; MDR1; Mdr1a; Mdr1b; Mdr3; p-170; P-GP; Pgp; Pgy-1; Pgy-3; PGY1; Pgy2; Pgy3

          View more View less

          UniProt ID: (Human) P08183, (Mouse) P21447, (Mouse) P06795

          View more View less

          Entrez Gene ID: (Dog) 403879, (Human) 5243, (Bovine) 281585, (Rat) 170913, (Mouse) 18671, (Mouse) 18669, (Rat) 24646

          View more View less

          Function(s)
          protein binding ATP binding xenobiotic-transporting ATPase activity efflux transmembrane transporter activity ATPase activity transmembrane transporter activity ubiquitin protein ligase binding ATPase activity, coupled to transmembrane movement of substances xenobiotic transporter activity carboxylic acid transmembrane transporter activity phosphatidylcholine-translocating ATPase activity phosphatidylethanolamine flippase activity ceramide floppase activity ATPase-coupled intramembrane lipid transporter activity floppase activity phosphatidylethanolamine floppase activity ABC-type transporter activity ATP-binding cassette (ABC) transporter
          Process(es)
          G2/M transition of mitotic cell cycle xenobiotic metabolic process response to xenobiotic stimulus phospholipid translocation terpenoid transport transmembrane transport transepithelial transport stem cell proliferation ceramide translocation export across plasma membrane transport across blood-brain barrier carboxylic acid transmembrane transport drug transmembrane export drug transport across blood-brain barrier regulation of chloride transport epidermal growth factor receptor signaling pathway heart development adult heart development female gonad development response to toxic substance gene expression proteasomal protein catabolic process neurogenesis exploration behavior aldosterone secretion xenobiotic transport cardiac muscle cell differentiation neural tissue regeneration tissue homeostasis protein acetylation establishment of endothelial blood-brain barrier neuron differentiation Sertoli cell barrier remodeling response to hypoxia placenta development lactation response to mycotoxin response to ionizing radiation response to lipopolysaccharide response to vitamin A response to genistein response to arsenic-containing substance response to steroid hormone cellular response to external biotic stimulus cellular response to tumor necrosis factor response to antineoplastic agent response to glycoside
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-xmwlv:80/100.66.76.16:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline