Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytokeratin Pan Type I Antibodies

          Invitrogen

          Cytokeratin Type I Monoclonal Antibody (AE-1)

          2 References
          View all (34) Cytokeratin Pan Type I antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cytokeratin Type I Monoclonal Antibody (AE-1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          MA1-35367

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:100
          -

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:150-1:200
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1:150-1:200
          -
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Human, Mouse, Rabbit, Rat

          Published species

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AE-1

          Immunogen

          Human epidermal keratin.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          250 µg

          Purification

          purified

          Storage buffer

          PBS

          Contains

          0.1% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2134400

          Product Specific Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          Target Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 40-kDa keratin intermediate filament; 56 kDa cytokeratin; Acidic Cytokeratin; alpha keratin; cell proliferation-inducing gene 46 protein; CK-10; CK-14; CK-15; CK-18; CK-19; CK-9; cytokeratin 10; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 9; cytokeratin endo B; cytokeratin VIB; cytokeratin VII; cytokeratin-10; cytokeratin-14; cytokeratin-15; cytokeratin-16; cytokeratin-18; cytokeratin-19; cytokeratin-6B; cytokeratin-7; cytokeratin-9; epidermal keratin 10; epidermal keratin type I; epidermal keratin VII; epidermolytic hyperkeratosis; keratosis palmaris et plantaris; focal non-epidermolytic palmoplantar keratoderma; HMWCK; intermediate filament; intermediate filament protein; intermediate filament protein K16; K10; K14; K15; K18; K19; K9; keratin; keratin 10 (epidermolytic hyperkeratosis); keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 14; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin 14-like; keratin 15; keratin 15, type I; keratin 16 homolog; keratin 16, type I; keratin 18, type I; keratin 19; keratin 19, type I; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 1, acidic, gene 9; keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin, type I cytoskeletal 59 kDa; keratin, type I, 40-kd; keratin-10; keratin-15, basic; keratin-15, beta; ORF; pan keratin; pankeratin; PIG46; spermatid perinuclear ring manchette protein K9; suprabasal cytokeratin 10; type I alpha-keratin 14; type I alpha-keratin 15; type I cytokeratin; type I cytoskeletal 15; type I cytoskeletal 9; type I keratin KA10; type I keratin Ka14; type I keratin KA15; type I keratin KA16; type I keratin KA19; unnamed protein product

          View more View less

          Gene Aliases: BCIE; BIE; CK-14; CK-16; CK-18; CK-19; CK-9; CK10; CK14; CK15; CK16; CK18; CK19; CYK18; D130054E02Rik; EBS1; EBS1A; EBS1B; EBS1C; EBS1D; EBS3; EBS4; EHK; EHK2; EHK2A; EHK2B; EndoC; EPPK; FNEPPK; GK-19; IHL; K10; K14; K15; K16; K18; K19; K1C1; K1CO; K1CP; K1CS; K9; Ka10; Ka14; Ka15; Ka16; Ka19; keratin-19; KPP; Krt-1.10; Krt-1.14; Krt-1.19; Krt1-10; Krt1-14; Krt1-15; Krt1-16; Krt1-18; Krt1-19; Krt1-9; Krt14; Krt14l; KRT15; KRT16A; KRT19; NEPPK; NFJ; PC1

          View more View less

          UniProt ID: (Bovine) P06394, (Human) P13645, (Human) P02533, (Chicken) Q6PVZ1, (Human) P19012, (Human) P08779, (Human) P05783, (Bovine) P08728, (Human) P08727, (Chicken) O93256, (Human) P35527, (Mouse) P02535, (Rat) Q6IFW6, (Mouse) Q61781, (Rat) Q6IFV1, (Mouse) Q61414, (Rat) Q6IFV3, (Mouse) Q9Z2K1, (Rat) Q5BJY9, (Mouse) P05784, (Mouse) P19001, (Rat) Q63279, (Mouse) Q6RHW0, (Rat) Q8CIS9

          View more View less

          Entrez Gene ID: (Bovine) 281888, (Chicken) 771977, (Rabbit) 100009029, (Human) 3858, (Chicken) 408040, (Human) 3861, (Bovine) 404111, (Chicken) 408039, (Rabbit) 100341120, (Bovine) 507464, (Human) 3866, (Human) 3868, (Rabbit) 100341376, (Rabbit) 100354174, (Chicken) 101749333, (Bovine) 506480, (Human) 3875, (Bovine) 514812, (Human) 3880, (Chicken) 395861, (Human) 3857, (Mouse) 16661, (Rat) 450225, (Mouse) 16664, (Rat) 287701, (Mouse) 16665, (Rat) 287700, (Mouse) 16666, (Rat) 303530, (Rat) 294853, (Mouse) 16668, (Mouse) 16669, (Rat) 360626, (Mouse) 107656, (Rat) 266717

          View more View less

          Function(s)
          structural molecule activity protein heterodimerization activity protein binding structural constituent of epidermis structural constituent of cytoskeleton keratin filament binding scaffold protein binding RNA binding structural constituent of muscle cadherin binding involved in cell-cell adhesion macromolecular complex binding cytoskeletal protein binding
          Process(es)
          Notch signaling pathway response to estrogen sarcomere organization cell differentiation involved in embryonic placenta development morphogenesis of an epithelium epithelial cell differentiation wound healing intermediate filament organization negative regulation of keratinocyte differentiation keratinocyte development epidermis development keratinocyte differentiation positive regulation of epidermis development protein heterotetramerization cornification cellular response to calcium ion spermatogenesis skin development inflammatory response cytoskeleton organization negative regulation of cell migration keratinization hair cycle innate immune response keratinocyte migration establishment of skin barrier response to fructose response to nutrient levels tumor necrosis factor-mediated signaling pathway Golgi to plasma membrane protein transport negative regulation of apoptotic process intermediate filament cytoskeleton organization hepatocyte differentiation mesenchymal stem cell differentiation extrinsic apoptotic signaling pathway hepatocyte apoptotic process cholangiocyte apoptotic process intermediate filament bundle assembly positive regulation of keratinocyte differentiation response to radiation response to zinc ion response to ionizing radiation negative regulation of epithelial cell differentiation biological_process anatomical structure morphogenesis cell-cell adhesion intermediate filament-based process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0