Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Insulin Antibodies

          Invitrogen

          Insulin Monoclonal Antibody (2D11-H5)

          1 Reference
          View all (85) Insulin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Insulin Monoclonal Antibody (2D11-H5)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Insulin Monoclonal Antibody (2D11-H5)

          Product Details

          MA1-35525

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Pig, Rabbit

          Published species

          Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2D11-H5

          Immunogen

          Purified pig insulin, conjugated to KLH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          100 µg

          Purification

          purified

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1076193

          Product Specific Information

          Insulin is a protein consisting of an α-chain of 21 amino acids and a β-chain of 30 amino acids and produced in the β-cells of the pancreas. 2D11-H5 is a specific insulin antibody as tested by ELISA and on human pancreatic tissue.

          Target Information

          Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ins; INSU1; INSU2; INSU3B; INSUA; INSUB; INSUC; insulin splice variant; insulin variant; INS-splice; Insulin-1; preproinsulin; proinsulin; unnamed protein product

          View more View less

          Gene Aliases: IDDM; IDDM1; IDDM2; ILPR; Ins-1; Ins-2; Ins2-rs1; InsII; IRDN; Mody; MODY10; Mody4; PNDM4

          View more View less

          UniProt ID: (Pig) P01315, (Human) P01308, (Mouse) P01325, (Mouse) P01326

          View more View less

          Entrez Gene ID: (Pig) 397415, (Bovine) 280829, (Rabbit) 100009181, (Human) 3630, (Mouse) 16333, (Mouse) 16334

          View more View less

          Function(s)
          protease binding insulin receptor binding insulin-like growth factor receptor binding hormone activity identical protein binding protein binding receptor agonist activity zinc ion binding growth factor
          Process(es)
          negative regulation of acute inflammatory response glucose metabolic process regulation of cellular amino acid metabolic process acute-phase response G-protein coupled receptor signaling pathway positive regulation of cell proliferation insulin receptor signaling pathway lipid biosynthetic process glycoprotein biosynthetic process positive regulation of nitric oxide mediated signal transduction positive regulation of glucose metabolic process positive regulation of phosphatidylinositol 3-kinase signaling lactate biosynthetic process regulation of transmembrane transporter activity positive regulation of cell migration positive regulation of protein autophosphorylation activation of protein kinase B activity negative regulation of NAD(P)H oxidase activity wound healing lipoprotein biosynthetic process negative regulation of protein catabolic process glucose homeostasis positive regulation of MAPK cascade negative regulation of gluconeogenesis positive regulation of fatty acid biosynthetic process positive regulation of glycogen biosynthetic process positive regulation of DNA replication negative regulation of glycogen catabolic process positive regulation of glycolytic process positive regulation of mitotic nuclear division negative regulation of proteolysis negative regulation of fatty acid metabolic process positive regulation of glucose import positive regulation of insulin receptor signaling pathway alpha-beta T cell activation negative regulation of protein secretion positive regulation of cytokine secretion positive regulation of peptidyl-tyrosine phosphorylation negative regulation of lipid catabolic process positive regulation of lipoprotein lipase activity positive regulation of NF-kappaB transcription factor activity positive regulation of protein kinase B signaling fatty acid homeostasis negative regulation of respiratory burst involved in inflammatory response positive regulation of respiratory burst positive regulation of protein localization to nucleus positive regulation of dendritic spine maintenance regulation of protein localization to plasma membrane negative regulation of reactive oxygen species biosynthetic process response to L-arginine neuron projection maintenance negative regulation of feeding behavior signal transduction feeding behavior protein secretion response to heat response to glucose positive regulation of gene expression response to nutrient levels positive regulation of insulin secretion response to food negative regulation of appetite positive regulation of mammary gland epithelial cell proliferation positive regulation of Rho protein signal transduction estradiol secretion negative regulation of apoptotic process glucose import in response to insulin stimulus positive regulation of protein secretion response to growth hormone positive regulation of peptide hormone secretion positive regulation of cell maturation negative regulation of lactation positive regulation of lactation positive regulation of blood circulation response to butyrate positive regulation of cytokine production regulation of transcription, DNA-templated cell-cell signaling negative regulation of gene expression positive regulation of neuron projection development positive regulation of cell growth regulation of protein localization nitric oxide-cGMP-mediated signaling pathway vasodilation positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of cell differentiation positive regulation of lipid biosynthetic process regulation of synaptic plasticity regulation of protein secretion cognition positive regulation of nitric-oxide synthase activity relaxation of vascular smooth muscle positive regulation of brown fat cell differentiation positive regulation of long-term synaptic potentiation cellular response to oxygen-containing compound negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway response to cytokine TORC1 signaling response to peptide hormone positive regulation of translation response to cAMP cellular response to glucose stimulus response to dimethyl sulfoxide glucose transmembrane transport negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process negative regulation of transcription from RNA polymerase II promoter ER overload response myoblast fusion regulation of gene expression myotube differentiation lipid catabolic process insulin processing regulation of phosphorylation intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress positive regulation of ERK1 and ERK2 cascade
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0