Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Myosin Skeletal Muscle Antibodies

          Invitrogen

          Myosin Skeletal Muscle Monoclonal Antibody (414)

          1 Reference
          View all (5) Myosin Skeletal Muscle antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Myosin Skeletal Muscle Monoclonal Antibody (414)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          Myosin Skeletal Muscle Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Myosin Skeletal Muscle Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Myosin Skeletal Muscle Antibody (MA1-35718) in WB

          Western blot was performed using Myosin Skeletal Muscle Monoclonal Antibody (414) (Product # MA1-35718) and a 223 kDa band corresponding to Myosin Skeletal Muscle was observed in RD, C2C12 differentiated to myotubes and L6 but not in HeLa, A549, MCF7 and PC12. Whole cell extracts (30 µg lysate) of RD (Lane 1), HeLa (Lane 2), A549 (Lane 3), MCF7 (Lane 4), C2C12 (Lane 5), C2C12 differentiated to myotubes (Lan... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Myosin Skeletal Muscle Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Myosin Skeletal Muscle Monoclonal Antibody (414)

          Product Details

          MA1-35718

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:5-1:10
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:5-1:10
          -

          Immunocytochemistry (ICC/IF)

          1:10
          -
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Rabbit

          Published species

          Rat

          Host/Isotype

          Mouse / IgM

          Class

          Monoclonal

          Type

          Antibody

          Clone

          414

          Immunogen

          Chicken skeletal muscle cell preparation
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          tissue culture supernatant

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2147020

          Product Specific Information

          Mab 414 reacts with myosin-containing elements in most cell types. Antigen localization: cytoplasm In Western blot experiments the antibody reacts with a 75kD part of the heavy chain (epitope location on a myosin-common sitechain), no reaction with light chains. It is not reactive with alpha-cardiac myosin. Positive control: muscle, myofibrils (chicken)

          Target Information

          Myosin is a major contractile protein which converts chemical energy into mechanical energy through the hydrolysis of ATP. Myosin is a hexameric protein composed of a pair of myosin heavy chains (MYH) and two pairs of nonidentical light chains. Myosin heavy chains are encoded by a multigene family. In mammals at least 10 different myosin heavy chain (MYH) isoforms have been described from striated, smooth, and nonmuscle cells. These isoforms show expression that is spatially and temporally regulated during development.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: epididymis luminal protein 71; fast 2a myosin heavy chain; inclusion body myopathy 3, autosomal dominant; MGC133384; myHC-2b; myHC-2x; myHC-IIx/d; myosin heavy chain 1; myosin heavy chain 2; myosin heavy chain 2A; myosin heavy chain 2b; myosin heavy chain 2X; myosin heavy chain 4; myosin heavy chain IIa; myosin heavy chain IIb; myosin heavy chain IIx/d; myosin heavy chain, skeletal muscle, adult; myosin heavy chain, skeletal muscle, adult 1; myosin heavy chain, skeletal muscle, adult 2; myosin heavy chain, skeletal muscle, fetal; myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin, heavy polypeptide 4, skeletal muscle; skeletal muscle myosin heavy chain II-a; type IIA myosin heavy chain; unnamed protein product

          View more View less

          Gene Aliases: A530084A17Rik; CMYO6; CMYP6; HEL71; IBM3; IId; IId/x; MdMs; MHC; MHC-2X/D; MHC2A; MHC2B; MHC2X/D; Minimsc; Minmus; MM; MYH-2B; MYH1; MYH2A; MYH2B; MYHa; MYHas8; MyHC-2A; MyHC-2B; MyHC-2x; MyHC-2X/D; MyHC-IIa; MyHC-IIb; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; Myhs-f; Myhs-f1; Myhs-f2; MYHSA1; MYHSA2; Myhsf; Myhsf1; Myhsf2

          View more View less

          UniProt ID: (Human) P12882, (Human) Q9UKX2, (Human) Q9Y623, (Rabbit) Q28641, (Mouse) Q5SX40, (Mouse) G3UW82, (Mouse) Q5SX39

          View more View less

          Entrez Gene ID: (Human) 4619, (Chicken) 427788, (Human) 4620, (Human) 4622, (Rabbit) 100125991, (Mouse) 17879, (Mouse) 17882, (Mouse) 17884

          View more View less

          Function(s)
          microfilament motor activity motor activity protein binding ATP binding actin filament binding calmodulin binding double-stranded RNA binding ATPase activity
          Process(es)
          muscle contraction sensory perception of sound muscle filament sliding actin filament-based movement ATP metabolic process response to muscle activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch